Merck
  • Mutations in STX1B, encoding a presynaptic protein, cause fever-associated epilepsy syndromes.
Mutations in STX1B, encoding a presynaptic protein, cause fever-associated epilepsy syndromes.
Nature genetics (2014-11-05)
Julian Schubert, Aleksandra Siekierska, Mélanie Langlois, Patrick May, Clément Huneau, Felicitas Becker, Hiltrud Muhle, Arvid Suls, Johannes R Lemke, Carolien G F de Kovel, Holger Thiele, Kathryn Konrad, Amit Kawalia, Mohammad R Toliat, Thomas Sander, Franz Rüschendorf, Almuth Caliebe, Inga Nagel, Bernard Kohl, Angela Kecskés, Maxime Jacmin, Katia Hardies, Sarah Weckhuysen, Erik Riesch, Thomas Dorn, Eva H Brilstra, Stephanie Baulac, Rikke S Møller, Helle Hjalgrim, Bobby P C Koeleman, Karin Jurkat-Rott, Frank Lehman-Horn, Jared C Roach, Gustavo Glusman, Leroy Hood, David J Galas, Benoit Martin, Peter A M de Witte, Saskia Biskup, Peter De Jonghe, Ingo Helbig, Rudi Balling, Peter Nürnberg, Alexander D Crawford, Camila V Esguerra, Yvonne G Weber, Holger Lerche
ABSTRACT
Febrile seizures affect 2-4% of all children and have a strong genetic component. Recurrent mutations in three main genes (SCN1A, SCN1B and GABRG2) have been identified that cause febrile seizures with or without epilepsy. Here we report the identification of mutations in STX1B, encoding syntaxin-1B, that are associated with both febrile seizures and epilepsy. Whole-exome sequencing in independent large pedigrees identified cosegregating STX1B mutations predicted to cause an early truncation or an in-frame insertion or deletion. Three additional nonsense or missense mutations and a de novo microdeletion encompassing STX1B were then identified in 449 familial or sporadic cases. Video and local field potential analyses of zebrafish larvae with antisense knockdown of stx1b showed seizure-like behavior and epileptiform discharges that were highly sensitive to increased temperature. Wild-type human syntaxin-1B but not a mutated protein rescued the effects of stx1b knockdown in zebrafish. Our results thus implicate STX1B and the presynaptic release machinery in fever-associated epilepsy syndromes.
Product Description
Supelco
Diethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diethylene glycol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Diethylene glycol, puriss. p.a., ≥99.0% (GC), colorless
USP
Diethylene glycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Diethylene glycol, BioUltra, ≥99.0% (GC)
Supelco
Diethylene glycol, analytical standard
Sigma-Aldrich
Diethylene glycol, ReagentPlus®, 99%