Merck
  • Multilocus sequence typing of Candida africana from patients with vulvovaginal candidiasis in New Delhi, India.
Multilocus sequence typing of Candida africana from patients with vulvovaginal candidiasis in New Delhi, India.
Mycoses (2014-04-05)
Cheshta Sharma, Sumathi Muralidhar, Jianping Xu, Jacques F Meis, Anuradha Chowdhary
ABSTRACT
We investigated the prevalence of vulvovaginal candidiasis due to C. africana in an STD clinic in India and analysed the genetic relatedness of these C. africana isolates with those outside India. A total of 283 germ-tube-positive yeasts were identified by VITEK2. Molecular characterisation of all isolates was carried out by hwp1-gene-specific PCR. Of 283 germ-tube-positive yeast isolates, four were identified as C. africana using hwp1-gene-specific PCR. All hwp1 PCR positive C. africana were subjected to antifungal susceptibility testing, ITS and D1/D2 region sequencing and were typed by using MLST approach. Similar to C. africana isolates from the United Kingdom and unlike those from Africa, the Indian C. africana grew at 42°C. Sequencing of eight gene fragments in MLST identified all four strains to have different genotypes not reported previously. Furthermore, though the Indian C. africana isolates were susceptible to most of the 14 tested antifungal drugs, differences in susceptibility were observed among the four strains. Our results indicate genetic and phenotypic heterogeneity among C. africana from different geographical regions. Due to lack of data on epidemiology and genetic variability of this under-reported yeast, more studies using molecular methods are warranted.
Product Description
Supelco
Clotrimazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
5-Fluorocytosine, nucleoside analog
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
Clotrimazole for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Ketoconazole, United States Pharmacopeia (USP) Reference Standard
USP
Clotrimazole, United States Pharmacopeia (USP) Reference Standard
USP
Miconazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Posaconazole, VETRANAL®, analytical standard
Sigma-Aldrich
Clotrimazole
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Miconazole, European Pharmacopoeia (EP) Reference Standard
Clotrimazole, European Pharmacopoeia (EP) Reference Standard
Flucytosine for system suitability, European Pharmacopoeia (EP) Reference Standard
Amphotericin B, European Pharmacopoeia (EP) Reference Standard
Fluconazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Fluconazole, European Pharmacopoeia (EP) Reference Standard
Voriconazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Fluconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Voriconazole, VETRANAL®, analytical standard
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
USP
Fluconazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Voriconazole, ≥98% (HPLC)
Flucytosine, European Pharmacopoeia (EP) Reference Standard