Merck
  • Simultaneous determination of metformin and vildagliptin in human plasma by a HILIC-MS/MS method.
Simultaneous determination of metformin and vildagliptin in human plasma by a HILIC-MS/MS method.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-07-13)
Roberto Pontarolo, Ana Carolina Gimenez, Thais Martins Guimarães de Francisco, Rômulo Pereira Ribeiro, Flávia Lada Degaut Pontes, João Cleverson Gasparetto
ABSTRACT
The objective of this work was to develop and validate a HILIC-MS/MS method for the simultaneous determination of metformin and vildagliptin in human plasma. Chromatographic separation was achieved using an Atlantis HILIC Silica 150-mm × 2.1-mm, 3-μm particle size column maintained at 40°C. The isocratic mobile phase consisted of 20% water and 80% acetonitrile/water solution 95:5 (v/v), containing both 0.1% formic acid and 3mM ammonium formate. The flow rate was maintained at 400 μL min(-1). Data from validation studies demonstrated that the new method is highly selective, sensitive (limits of detection <1.5 ng mL(-1)) and free of matrix and residual effects. The new method was also precise (RSD<9.0%), accurate (RE<11.2%) and linear (r ≥ 0.99) over the ranges of 5-500 ng mL(-1) for each compound. The developed method was successfully applied to determine metformin and vildagliptin in plasma volunteers who orally received a single dose of metformin (850 mg), vildagliptin (50mg) or drug association (metformin 850 mg+vildagliptin 50mg). The new method can thus also be used as a tool for the clinical monitoring of metformin and vildagliptin.
Product Description
Supelco
Metformin hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
USP
Metformin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Metformin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Pyrantel embonate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium formate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
1,1-Dimethylbiguanide hydrochloride, 97%
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Supelco
Pyrantel pamoate, VETRANAL®, analytical standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Supelco
Metformin Related Compound B, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid, reagent grade, ≥95%