Merck
  • Unimolecular reaction chemistry of a charge-tagged beta-hydroxyperoxyl radical.
Unimolecular reaction chemistry of a charge-tagged beta-hydroxyperoxyl radical.
Physical chemistry chemical physics : PCCP (2014-10-18)
Sui So, Benjamin B Kirk, Adam J Trevitt, Uta Wille, Stephen J Blanksby, Gabriel da Silva
ABSTRACT
β-Hydroxyperoxyl radicals are formed during atmospheric oxidation of unsaturated volatile organic compounds such as isoprene. They are intermediates in the combustion of alcohols. In these environments the unimolecular isomerization and decomposition of β-hydroxyperoxyl radicals may be of importance, either through chemical or thermal activation. We have used ion-trap mass spectrometry to generate the distonic charge-tagged β-hydroxyalkyl radical anion, ˙CH2C(OH)(CH3)CH2C(O)O(-), and investigated its subsequent reaction with O2 in the gas phase under conditions that are devoid of complicating radical-radical reactions. Quantum chemical calculations and master equation/RRKM theory modeling are used to rationalize the results and discern a reaction mechanism. Reaction is found to proceed via initial hydrogen abstraction from the γ-methylene group and from the β-hydroxyl group, with both reaction channels eventually forming isobaric product ions due to loss of either ˙OH + HCHO or ˙OH + CO2. Isotope labeling studies confirm that a 1,5-hydrogen shift from the β-hydroxyl functionality results in a hydroperoxyalkoxyl radical intermediate that can undergo further unimolecular dissociations. Furthermore, this study confirms that the facile decomposition of β-hydroxyperoxyl radicals can yield ˙OH in the gas phase.
Sigma-Aldrich
Oxygen-18O2, 99 atom % 18O, 99% (CP)
Sigma-Aldrich
Oxygen-18O2, 10 atom % 18O (random)
Sigma-Aldrich
3-Hydroxy-3-methylglutaric acid, ≥95%
Sigma-Aldrich
Oxygen-18O2, 97 atom % 18O
Sigma-Aldrich
Oxygen-18O2, 90 atom % 18O
Sigma-Aldrich
Oxygen-18O2, 50 atom % 18O (random)
Sigma-Aldrich
Oxygen-18O2, 25 atom % 18O (random)
Sigma-Aldrich
Deuterium, 99.9 atom % D
Sigma-Aldrich
Deuterium, 99.96 atom % D
Sigma-Aldrich
Deuterium, 99.8 atom % D
Sigma-Aldrich
Deuterium hydride, extent of labeling: 96 mol% DH, 98 atom % D