Merck
RASA3 is a critical inhibitor of RAP1-dependent platelet activation.
The Journal of clinical investigation (2015-02-24)
Lucia Stefanini, David S Paul, Raymond F Robledo, E Ricky Chan, Todd M Getz, Robert A Campbell, Daniel O Kechele, Caterina Casari, Raymond Piatt, Kathleen M Caron, Nigel Mackman, Andrew S Weyrich, Matthew C Parrott, Yacine Boulaftali, Mark D Adams, Luanne L Peters, Wolfgang Bergmeier
ABSTRACT
The small GTPase RAP1 is critical for platelet activation and thrombus formation. RAP1 activity in platelets is controlled by the GEF CalDAG-GEFI and an unknown regulator that operates downstream of the adenosine diphosphate (ADP) receptor, P2Y12, a target of antithrombotic therapy. Here, we provide evidence that the GAP, RASA3, inhibits platelet activation and provides a link between P2Y12 and activation of the RAP1 signaling pathway. In mice, reduced expression of RASA3 led to premature platelet activation and markedly reduced the life span of circulating platelets. The increased platelet turnover and the resulting thrombocytopenia were reversed by concomitant deletion of the gene encoding CalDAG-GEFI. Rasa3 mutant platelets were hyperresponsive to agonist stimulation, both in vitro and in vivo. Moreover, activation of Rasa3 mutant platelets occurred independently of ADP feedback signaling and was insensitive to inhibitors of P2Y12 or PI3 kinase. Together, our results indicate that RASA3 ensures that circulating platelets remain quiescent by restraining CalDAG-GEFI/RAP1 signaling and suggest that P2Y12 signaling is required to inhibit RASA3 and enable sustained RAP1-dependent platelet activation and thrombus formation at sites of vascular injury. These findings provide insight into the antithrombotic effect of P2Y12 inhibitors and may lead to improved diagnosis and treatment of platelet-related disorders.
Product Description
USP
Clopidogrel bisulfate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2,3-Diaminonaphthalene, BioReagent, suitable for fluorescence, ≥98.0% (HPLC)
Sigma-Aldrich
2,3-Diaminonaphthalene, ≥95% (HPLC), powder
Sigma-Aldrich
2-Methylthioadenosine 5′-monophosphate triethylammonium salt hydrate, solid, ≥98% (HPLC)
Sigma-Aldrich
N-Nitroso-N-ethylurea, ISOPAC®
Clopidogrel hydrogen sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
N-Nitroso-N-ethylurea, Bulk package
Supelco
Clopidogrel Bisulfate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Copper(II) chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Copper(II) chloride, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) chloride, 97%
Sigma-Aldrich
Copper(II) chloride, SAJ first grade, ≥98.0%
Sigma-Aldrich
(S)-(+)-Clopidogrel hydrogensulfate, ≥98% (HPLC)
Sigma-Aldrich
Copper(II) chloride, powder, 99%
Clopidogrel for system suitability, European Pharmacopoeia (EP) Reference Standard