Merck
  • A Huaier polysaccharide inhibits hepatocellular carcinoma growth and metastasis.
A Huaier polysaccharide inhibits hepatocellular carcinoma growth and metastasis.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-11-07)
Cong Li, Xia Wu, Honghai Zhang, Gengxia Yang, Meijun Hao, Shoupeng Sheng, Yu Sun, Jiang Long, Caixia Hu, Xicai Sun, Li Li, Jiasheng Zheng
ABSTRACT
This study was carried out to evaluate the effects of a Huaier polysaccharide (TP-1) on the tumor growth and immune function in hepatocellular carcinoma (HCC) H22-based mouse in vivo. Results showed that TP-1 was capable of repressing transplanted H22 solid hepatic tumor cell growth in vivo, prolonging the live time of mice bearing ascetic H22 tumors, and repressing the pulmonary metastasis of H22-bearing mice. Moreover, the relative weight of immune organ (spleen and thymus) and lymphocyte proliferation were improved after TP-1 treatment. Furthermore, the treatment with TP-1 could promote immune-stimulating serum cytokines, such as IL-2 and IFN-γ, but inhibit immune-suppressing serum cytokines IL-10 secretion in H22-bearing mice. Besides, the percentage of CD4+ T cells and NK cells was increased, whereas the number of CD8+ T cells decreased in tumor-bearing mice following TP-1 administration. In addition, this compound displayed little toxic effects to major organ of tumor-bearing mice at the therapeutic dose, such as the liver and kidney. This experimental finding suggested that TP-1 exhibited prominent antitumor activities in vivo via enhancement of host immune system function in H22 tumor-bearing mice. This product could be developed individually as a safe and potent biological response modifier for HCC therapy.
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
3-Methyl-1-butanol, SAJ first grade, ≥96.0%
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
3-Methyl-1-butanol, JIS special grade, ≥98.0%
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
3-Methyl-1-butanol, anhydrous, ≥99%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
3-Methyl-1-butanol, analytical standard
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
3-Methyl-1-butanol, reagent grade, 98%