Merck
  • Development and optimization of a method for analyzing biodiesel mixtures with non-aqueous reversed phase liquid chromatography.
Development and optimization of a method for analyzing biodiesel mixtures with non-aqueous reversed phase liquid chromatography.
Journal of chromatography. A (2008-03-18)
G Di Nicola, M Pacetti, F Polonara, G Santori, R Stryjek
ABSTRACT
Biodiesel (a mixture of fatty acid esters) is normally analyzed using gas chromatography/flame ionization detection, as specified by the ASTM D6584 and EN14105 standards. This paper proposes a binary gradient method for analyzing biodiesel mixtures using non-aqueous reverse phase HPLC with a UV detector capable of overcoming the drawbacks of the gas chromatographic technique normally used. The new analytical method was developed by means of a statistical sensitivity analysis applied to the main parameters influencing the recording, using the full factorial design method combined with the Yates algorithm and the steepest ascent optimization procedure. The present study shows the influence of the main biodiesel mixture separation analysis parameters. The resulting tool proved valid for analyzing not only biodiesel but also any traces of unreacted oil.
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%