Merck
  • Structure-activity relationship: analyses of p-glycoprotein substrates and inhibitors.
Structure-activity relationship: analyses of p-glycoprotein substrates and inhibitors.
Journal of clinical pharmacy and therapeutics (2003-06-11)
R B Wang, C L Kuo, L L Lien, E J Lien
ABSTRACT
A large number of structurally and functionally diverse compounds act as substrates or modulators of p-glycoprotein (p-gp). Some of them possess multiple drug resistance (MDR)-reversing activity, but only a small number of them have entered clinical study. In order to uncover the factors which exert a significant impact on the interaction between substrates/modulators and p-gp, we have performed structure-activity relationship (SAR) analyses, including molecular modelling, two-dimensional (2D) and three-dimensional (3D) parameter-frame-setting analysis, quantitative structure activity relationship (QSAR) analysis among substrates/modulators, as well as clinically promising MDR-reversing agents. The physicochemical parameters C log P, CMR and all regression equations were derived by using C log P version 4.0 and the latest CQSAR software, respectively. Molecular modelling and all other parameter calculations were performed by using HyperChem version 5.0 program, after geometry optimization and energy minimization using the AM1 semiempirical method. SAR analyses indicate that MDR reversal activity is correlated with the lipophilicity (C log P), molecular weight (log Mw), longest chain (Nlc) of the molecule and the energy of the highest occupied orbital (Ehomo). In addition, the presence of a basic tertiary nitrogen atom in the structure is also an important contributor to p-gp inhibitory activity. Some separation in space is achieved for different subsets of p-gp substrates and inhibitors using Nlc, C log P and Ehomo as three independent parameters in the 3D-parameter-frame setting. A highly effective p-gp modulator candidate should possess a log P value of 2.92 or higher, 18-atom-long or longer molecular axis, and a high Ehomo value, as well as at least one tertiary basic nitrogen atom. The results obtained may be useful in explaining drug-p-gp interactions for different compounds, including drug interactions and the development of new MDR chemosensitizers.
Sigma-Aldrich
SB-MDR1-Hi5-PREDEASY-ATPase kit, membrane based kit for Human MDR1 ATPase Transport Assays
Sigma-Aldrich
SB-ratMdr1b-Sf9-PREDEASY-ATPase kit, membrane based kit for Rat Mdr1b ATPase Transport Assays