Merck
  • Determination of iodide in seawater and urine by size exclusion chromatography with iodine-starch complex.
Determination of iodide in seawater and urine by size exclusion chromatography with iodine-starch complex.
Journal of chromatography. A (2001-06-16)
H B Li, F Chen, X R Xu
ABSTRACT
A novel method for the determination of iodide by size exclusion chromatography was established. The method was simple and highly sensitive with good precision. Iodide was converted to iodine, then sequestered with starch, and separated from the matrix using a Shim-pack DIOL-150 (250 x 7.9 mm) size exclusion column with methanol-0.01 mol l(-1) aqueous phosphoric acid (10:90, v/v) as mobile phase at 1.2 ml min(-1) and UV detection at 224 nm. The calibration graph was linear from 1.0 ng ml(-1) to 100.0 ng ml(-1) for iodide with a correlation coefficient of 0.9992 (n=6). The detection limit was 0.2 ng ml(-1). The method was successfully applied to the determination of iodide in seawater and urine. The recovery was from 92% to 103% and the relative standard deviation was in the range of 1.5% to 3.7%.
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Pricing and availability is not currently available.