Merck
  • Protective effects of novel metal-nonoates on the cellular components of the vascular system.
Protective effects of novel metal-nonoates on the cellular components of the vascular system.
The Journal of pharmacology and experimental therapeutics (2014-09-23)
Martina Monti, Raffaella Solito, Luca Puccetti, Luca Pasotti, Riccardo Roggeri, Enrico Monzani, Luigi Casella, Lucia Morbidelli
ABSTRACT
At the cardiovascular level, nitric oxide (NO) controls smooth muscle functions, maintains vascular integrity, and exerts an antihypertensive effect. Metal-nonoates are a recently discovered class of NO donors, with NO release modulated through the complexation of the N-aminoethylpiperazine N-diazeniumdiolate ligand to metal ions, and thus representing a significant innovation with respect to the drugs traditionally used. In this study, we characterized the vascular protective effects of the most effective compound of this class, Ni(PipNONO)Cl, compared with the commercial N-diazeniumdiolate group derivate, diethylenetriamine/nitric oxide (DETA/NO). Ni(PipNONO)Cl induced a concentration-dependent relaxation of precontracted rat aortic rings. The ED50 was 0.67 µM, compared with 4.3 µM obtained with DETA/NO. When tested on cultured microvascular endothelial cells, Ni(PipNONO)Cl exerted a protective effect on the endothelium, promoting cell proliferation and survival in the picomolar range. The administration of Ni(PipNONO)Cl to vascular smooth muscle cells reduced the cell number, promoting their apoptosis at a high concentration (10 µM). Inhibition of smooth muscle cell migration, a hallmark of atherosclerosis, was accompanied by cytoskeletal rearrangement and loss of lamellipodia. When added to isolated platelets, Ni(PipNONO)Cl significantly reduced ADP-induced aggregation. Since atherosclerosis is accompanied by an inflammatory environment, cultured endothelial cells were exposed to interleukin (IL)-1β. In the presence of IL-1β, Ni(PipNONO)Cl inhibited cyclooxygenase-2 and inducible nitric oxide synthase upregulation, and reduced endothelial permeability and the platelet and monocyte adhesion markers CD31 and CD40 at the plasma membrane. Overall, these data indicate that Ni(PipNONO)Cl exerts vascular protective effects relevant for vascular dysfunction and prevention of atherosclerosis and thrombosis.
Product Description
SAFC
BIS-TRIS
Sigma-Aldrich
Anti-Tubulin antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate, ≥98% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
SAFC
BIS-TRIS
Sigma-Aldrich
3-[(1R)-1-Hydroxy-2-(methylamino)ethyl]phenol, AldrichCPR
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate sodium salt, ≥99% (HPLC), powder
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-eNOS/NOS III (Thr495) Antibody, rabbit monoclonal, culture supernatant, Upstate®
Phenylephrine, European Pharmacopoeia (EP) Reference Standard