Merck
  • Redox-sensitive gene-regulatory events controlling aberrant matrix metalloproteinase-1 expression.
Redox-sensitive gene-regulatory events controlling aberrant matrix metalloproteinase-1 expression.
Free radical biology & medicine (2014-06-29)
Toni R Bartling, Sita Subbaram, Ryan R Clark, Akshaya Chandrasekaran, Supriya Kar, J Andres Melendez
ABSTRACT
Aberrant matrix metalloproteinase-1 (MMP-1) expression contributes to the pathogenesis of many degenerative disease processes that are associated with increased oxidative damage or stress. We and others have established that shifts in steady-state H2O2 production resulting from enforced antioxidant gene expression, senescence, or UV irradiation control MMP-1 expression. Here we establish that histone deacetylase-2 (HDAC2) protein levels and its occupancy of the MMP-1 promoter are decreased in response to enforced manganese superoxide dismutase (Sod2) expression. Inhibition of HDAC activity further accentuates the redox-dependent expression of MMP-1. Sod2-dependent decreases in HDAC2 are associated with increases in a proteasome-sensitive pool of ubiquitinylated HDAC2 and MMP-1-specific histone H3 acetylation. Sod2 overexpression also enhanced recruitment of Ets-1, c-Jun, c-Fos, and the histone acetyltransferase PCAF to the distal and proximal regions of the MMP-1 promoter. Furthermore, the Sod2-dependent expression of MMP-1 can be reversed by silencing the transcriptional activator c-Jun. All of the above Sod2-dependent alterations are largely reversed by catalase coexpression, indicating that the redox control of MMP-1 is H2O2-dependent. These findings identify a novel redox regulation of MMP-1 transcription that involves site-specific promoter recruitment of both activating factors and chromatin-modifying enzymes, which converge to maximally drive MMP-1 gene expression.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
3-Amino-1,2,4-triazole, ≥95% (TLC)
Supelco
Amitrol, PESTANAL®, analytical standard
Sigma-Aldrich
EZ-Magna ChIP® A/G Chromatin Immunoprecipitation Kit, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A/G beads. Control primers included.
Sigma-Aldrich
Anti-Histone H3 Antibody, 0.5 mg/mL, Upstate®
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
4H-1,2,4-Triazol-3-amine, AldrichCPR
Trapidil impurity B, European Pharmacopoeia (EP) Reference Standard