Merck
  • Xenopus laevis FGF receptor substrate 3 (XFrs3) is important for eye development and mediates Pax6 expression in lens placode through its Shp2-binding sites.
Xenopus laevis FGF receptor substrate 3 (XFrs3) is important for eye development and mediates Pax6 expression in lens placode through its Shp2-binding sites.
Developmental biology (2014-12-03)
Yeon-Jin Kim, Minjin Bahn, Yong Hwan Kim, Jee-Yoon Shin, Seon-Woo Cheong, Bong-Gun Ju, Won-Sun Kim, Chang-Yeol Yeo
ABSTRACT
Members of the fibroblast growth factor (FGF) family play important roles during various developmental processes including eye development. FRS (FGF receptor substrate) proteins bind to FGFR and serve as adapters for coordinated assembly of multi-protein complexes involved in Ras/MAPK and PI3 kinase/Akt pathways. Here, we identified Xenopus laevis Frs3 (XFrs3), a homolog of vertebrate Frs3, and investigated its roles during embryogenesis. XFrs3 is expressed maternally and zygotically with specific expression patterns throughout the early development. Knockdown of XFrs3 using a specific antisense morpholino oligonucleotide (MO) caused reduction of Pax6 expression in the lens placode, and defects in the eye ranging from microphthalmia to anophthalmia. XFrs3 MO-induced defects were alleviated by wild type XFrs3 or a mutant XFrs3 (XFrs3-4YF), in which the putative tyrosine phosphorylation sites served as Grb2-binding sites are mutated. However, another XFrs3 mutant (XFrs3-2YF), in which the putative Shp2-binding sites are mutated, could not rescue the defects of XFrs3 morphants. In addition, we found that XFrs3 is important for FGF or IGF-induced ERK activation in ectodermal tissue. Taken together, our results suggest that signaling through Shp2-binding sites of XFrs3 is necessary for the eye development in Xenopus laevis.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium Fluoride Solution
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 1 M
USP
L-Tyrosine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture