Merck
  • Characterization of photo-transformation products of the antibiotic drug Ciprofloxacin with liquid chromatography-tandem mass spectrometry in combination with accurate mass determination using an LTQ-Orbitrap.
Characterization of photo-transformation products of the antibiotic drug Ciprofloxacin with liquid chromatography-tandem mass spectrometry in combination with accurate mass determination using an LTQ-Orbitrap.
Chemosphere (2014-03-19)
Tarek Haddad, Klaus Kümmerer
ABSTRACT
The presence of pharmaceuticals, especially antibiotics, in the aquatic environment is of growing concern. Several studies have been carried out on the occurrence and environmental risk of these compounds. Ciprofloxacin (CIP), a broad-spectrum anti-microbial second-generation fluoroquinolone, is widely used in human and veterinary medicine. In this work, photo-degradation of CIP in aqueous solution using UV and xenon lamps was studied. The transformation products (TPs), created from CIP, were initially analyzed by an ion trap in the MS, MS/MS and MS(3) modes. These data were used to clarify the structures of the degradation products. Furthermore, the proposed products were confirmed by accurate mass measurement and empirical formula calculation for the molecular ions of TPs using LTQ-Orbitrap XL mass spectrometer. The degree of mineralization, the abundance of detected TPs and degradation pathways were determined. Eleven TPs were detected in the present study. TP1, which was never detected before, was structurally characterized in this work. All TPs still retained the core quinolone structure, which is responsible for the biological activity. As mineralization of CIP and its transformation products did not happen, the formation of stable TPs can be expected in waste water treatment and in surface water with further follow-up problems.
Product Description
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Ciprofloxacin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Ciprofloxacin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Ciprofloxacin HCl, Pharmaceutical Secondary Standard; Certified Reference Material