Merck
  • Angiographic and echocardiographic correlates of suitable septal perforators for alcohol septal ablation in hypertrophic obstructive cardiomyopathy.
Angiographic and echocardiographic correlates of suitable septal perforators for alcohol septal ablation in hypertrophic obstructive cardiomyopathy.
The Canadian journal of cardiology (2014-07-30)
William Chan, Lynne Williams, Mark A Kotowycz, Anna Woo, Harry Rakowski, Leonard Schwartz, Christopher B Overgaard
ABSTRACT
Myocardial contrast echocardiography during angiography is critical in identifying appropriate septal perforator(s) for alcohol septal ablation (ASA) in patients with hypertrophic obstructive cardiomyopathy. We evaluated whether there were other angiographic and/or echocardiographic markers that might identify patients who are anatomically suitable for ASA. We performed quantitative coronary angiographic analysis and echocardiographic assessment on 74 patients referred for ASA from January 2004 to July 2012 at the Peter Munk Cardiac Centre, University Health Network, Toronto, Ontario, Canada. Patients who proceeded to ASA were compared with those in whom ASA was aborted. Of the 74 patients referred for ASA, 63 proceeded to ASA and in 11 patients ASA was aborted because of various anatomic and technical reasons. There were no clinically significant differences observed in quantitative angiographic and echocardiographic measurements between the 2 groups. The ratio of ostial left main (LM) to ablated septal distance on angiography vs the basal septum to the septum area where the mitral valve contacted the septum because of systolic anterior motion (SAM) was 1.53. In the whole cohort, a significant correlation was observed between the ostial LM to the target septal distance and the distance from basal septum to SAM-septal contact point on echocardiography (r = 0.39; P = 0.008). A stronger correlation was evident when analysis was restricted to patients undergoing ASA only (r = 0.44; P = 0.006). Echocardiographic and angiographic assessments of the distance between the basal septum to SAM-septal contact point and ostial LM to the target septal distance might be useful in preprocedural selection of the appropriate septal perforator for ASA.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Reagent Alcohol, reagent grade
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Reagent Alcohol, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, denatured, reagent grade
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology