Merck
  • Collateral sensitivity of resistant MRP1-overexpressing cells to flavonoids and derivatives through GSH efflux.
Collateral sensitivity of resistant MRP1-overexpressing cells to flavonoids and derivatives through GSH efflux.
Biochemical pharmacology (2014-05-31)
Doriane Lorendeau, Lauriane Dury, Estelle Genoux-Bastide, Florine Lecerf-Schmidt, Claudia Simões-Pires, Pierre-Alain Carrupt, Raphaël Terreux, Sandrine Magnard, Attilio Di Pietro, Ahcène Boumendjel, Hélène Baubichon-Cortay
ABSTRACT
The multidrug resistance protein 1 (MRP1) is involved in multidrug resistance of cancer cells by mediating drug efflux out of cells, often in co-transport with glutathione (GSH). GSH efflux mediated by MRP1 can be stimulated by verapamil. In cells overexpressing MRP1, we have previously shown that verapamil induced a huge intracellular GSH depletion which triggered apoptosis of the cells. That phenomenon takes place in the more global anticancer strategy called "collateral sensitivity" and could be exploited to eradicate some chemoresistant cancer cells. Seeking alternative compounds to verapamil, we screened a library of natural flavonoids and synthetic derivatives. A large number of these compounds stimulate MRP1-mediated GSH efflux and the most active ones have been evaluated for their cytotoxic effect on MRP1-overexpressing cells versus parental cells. Interestingly, some are highly and selectively cytotoxic for MRP1-cells, leading them to apoptosis. However, some others do not exhibit any cytotoxicity while promoting a strong GSH efflux, indicating that GSH efflux is necessary but not sufficient for MRP1-cells apoptosis. In support to this hypothesis, structure activity relationships show that the absence of a hydroxyl group at position 3 of the flavonoid C ring is an absolute requirement for induction of MRP1-cells death, but is not for GSH efflux stimulation. Chrysin (compound 8) and its derivatives, compounds 11 and 22, exhibit a high selectivity toward MRP1-cells with a IC₅₀ value of 4.1 μM for compound 11 and 4.9 μM for chrysin and compound 22, making them among the best described selective killer compounds of multidrug ABC transporter-overexpressing cells.
Product Description
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzaldehyde, SAJ special grade, ≥98.0%
Sigma-Aldrich
2′-Hydroxy-4′,6′-dimethoxyacetophenone, 97%
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Supelco
Benzaldehyde, analytical standard
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzaldehyde, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Boron tribromide, ≥99.99%
Sigma-Aldrich
Boron tribromide, ReagentPlus®, ≥99%
Sigma-Aldrich
Boron tribromide, ReagentPlus®, 99.9%
Sigma-Aldrich
Boron tribromide solution, 1.0 M in hexanes
Sigma-Aldrich
Boron tribromide solution, 1.0 M in methylene chloride
Sigma-Aldrich
Boron tribromide solution, 1.0 M in heptane
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Colchicine, BioReagent, suitable for plant cell culture, ≥95% (HPLC)
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Colchicine, (European Pharmacopoeia (EP) Reference Standard)