Merck
  • [Mast cell activation disease: a concise practical guide for diagnostic workup and therapeutic options].
[Mast cell activation disease: a concise practical guide for diagnostic workup and therapeutic options].
Deutsche medizinische Wochenschrift (1946) (2014-05-08)
G J Molderings, J Homann, S Brettner, M Raithel, T Frieling
ABSTRACT
In the present paper clinical phenotypes, pathogenetic relationships, and diagnostic algorithms as well as therapeutic concepts of/for systemic mast cell activation disease are reviewed. The reader should be able to recognize and diagnose a systemic mast cell activation disease, as well as to counsel a personalized drug therapy. In the case of chronic multisystem polymorbidity systemic mast cell activation disease should be considered as a differential diagnosis at an early stage. In most cases, specific, little invasive investigations allow diagnosing the disease and, hence, an appropriate therapy can be initiated.
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
(+)-Sodium L-ascorbate, SAJ special grade, ≥99.0%
Sodium ascorbate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
Ketotifen fumarate salt
Ketotifen for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Ketotifen hydrogen fumarate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ranitidine hydrochloride, solid
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Ranitidine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ranitidine hydrochloride, United States Pharmacopeia (USP) Reference Standard