Merck
  • The chronic toxicity of ZnO nanoparticles and ZnCl2 to Daphnia magna and the use of different methods to assess nanoparticle aggregation and dissolution.
The chronic toxicity of ZnO nanoparticles and ZnCl2 to Daphnia magna and the use of different methods to assess nanoparticle aggregation and dissolution.
Nanotoxicology (2013-07-11)
Nathalie Adam, Claudia Schmitt, Josep Galceran, Encarna Companys, Alexander Vakurov, Rachel Wallace, Dries Knapen, Ronny Blust
ABSTRACT
In this study, the effect of ZnO nanoparticles and ZnCl2 on growth, reproduction and accumulation of zinc in Daphnia magna was determined in a 21-day chronic toxicity test. A variety of techniques were used to distinguish the free zinc ion, dissolved, nanoparticle and aggregated zinc fraction in the Daphnia test medium. The results showed similar chronic effects on growth, reproduction and accumulation for the ZnO nanoparticles (EC10, 20, 50 reproduction: 0.030, 0.049, 0.112 mg Zn/l) and the ZnCl2 (EC10, 20, 50 reproduction: 0.014, 0.027, 0.082 mg Zn/l). A large fraction of the nanoparticles rapidly dissolved after introduction in the exposure medium. Aggregation of nanoparticles was also observed but within 48 h of exposure most of these ZnO aggregates were dissolved. Based on the combined dissolution kinetics and toxicity results, it can be concluded that the toxicological effects of ZnO nanoparticles at the chronic level can be largely attributed to the dissolved fraction rather than the nanoparticles or initially formed aggregates.
Sigma-Aldrich
Zinc chloride solution, 1.9 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Zinc chloride, JIS special grade, ≥98.0%
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Zinc chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Zinc chloride, 99.999% trace metals basis
Sigma-Aldrich
Zinc chloride, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Zinc chloride 0.1 M solution
Sigma-Aldrich
Zinc chloride, SAJ first grade, ≥95.0%
Sigma-Aldrich
Zinc chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
Sigma-Aldrich
Zinc chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98-100.5%
Sigma-Aldrich
Zinc chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Zinc chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Zinc chloride solution, 0.5 M in THF
Sigma-Aldrich
Zinc chloride, reagent grade, ≥98%
Sigma-Aldrich
Zinc chloride, ACS reagent, ≥97%
Sigma-Aldrich
Zinc oxide, 99.999% trace metals basis
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sigma-Aldrich
Zinc oxide, 99.99% trace metals basis
Sigma-Aldrich
Zinc oxide, nanopowder, <100 nm particle size
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Zinc oxide, nanowires, Length 1 um
Sigma-Aldrich
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%
Sigma-Aldrich
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
Supelco
Zinc oxide, analytical standard