Merck
Cerium methods for light and electron microscopical histochemistry.
Journal of microscopy (1993-07-01)
C J Van Noorden, W M Frederiks
ABSTRACT
Methods based on the use of cerium to detect the activity of oxidases and phosphatases are rapidly expanding. Both classes of enzymes can be demonstrated with cerium at the electron and light microscopical level. The in situ detection of H2O2 production with cerium is another application that has great potential, particularly in experimental pathological research. The fine precipitate of the cerium-containing final reaction product, cerium perhydroxide or cerium phosphate, enables a very precise localization. With such techniques, important advances have been made in cell biology, such as the discovery of new organelles, functional subcompartmentization of peroxisomes, tubular lysosomes and the elucidation of the function of extracellular ATPases. At the light microscopical level, the activity of enzymes can be quantified in situ because the production of final reaction product in the cerium methods is proportional to enzyme activity in tissue sections or cells. Cerium precipitates have strong reflectance properties and this enables their use in confocal scanning laser microscopy. In the present review, the principles of cerium methods are outlined and applications in cell biology and pathology are discussed.
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Cerium, chips
Cerium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Cerium, wire reel, 25mm, diameter 1.0mm, hard, 99.9%
Cerium, foil, 50x50mm, thickness 0.1mm, as rolled, 99.9%
Cerium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Cerium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.9%
Cerium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.9%
Cerium, rod, 100mm, diameter 6.35mm, cast, 99.9%
Cerium, rod, 100mm, diameter 12.7mm, cast, 99.9%
Cerium, rod, 50mm, diameter 12.7mm, cast, 99.9%
Cerium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%
Cerium, rod, 50mm, diameter 6.35mm, cast, 99.9%