Merck
Recent advances in homogeneous nickel catalysis.
Nature (2014-05-16)
Sarah Z Tasker, Eric A Standley, Timothy F Jamison
ABSTRACT
Tremendous advances have been made in nickel catalysis over the past decade. Several key properties of nickel, such as facile oxidative addition and ready access to multiple oxidation states, have allowed the development of a broad range of innovative reactions. In recent years, these properties have been increasingly understood and used to perform transformations long considered exceptionally challenging. Here we discuss some of the most recent and significant developments in homogeneous nickel catalysis, with an emphasis on both synthetic outcome and mechanism.
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Nickel, foil, 50x50mm, thickness 0.3mm, as rolled, 99.999%
Nickel, foil, 25x25mm, thickness 2.0mm, annealed, 99%
Nickel, foil, 10mm disks, thickness 0.025mm, as rolled, 99.99+%
Nickel, tube, 500mm, outside diameter 1.8mm, inside diameter 1.6mm, wall thickness 0.1mm, hard, 99.5%
Nickel, foil, 25x25mm, thickness 0.125mm, as rolled, 99.999%
Nickel, pellets, 50g, max. size 20mm, 99.99+%
Nickel, foil, 0.5m coil, thickness 0.15mm, coil width 12mm, annealed, 99%
Nickel, microfoil, disks, 25mm, thinness 0.5μm, specific density 438.8μg/cm2, permanent mylar 3.5μm support, 99.99+%
Nickel, foil, 50x50mm, thickness 1.0mm, annealed, 99%
Nickel, foil, 2m coil, thickness 0.025mm, coil width 600mm, as rolled, 99%
Nickel, tube, 500mm, outside diameter 3.25mm, inside diameter 3.05mm, wall thickness 0.1mm, hard, 99.5%
Nickel, foil, 10mm disks, thickness 0.05mm, annealed, 99%
Nickel, wire reel, 10m, diameter 0.125mm, annealed, 99.98%
Nickel, rod, 1000mm, diameter 3.0mm, 99%
Nickel, mesh, 30x30mm, nominal aperture 0.34mm, thickness 0.025mm, wire diameter 0.041mm, 70 wires/inch, open area 80%, electro-formed mesh, 99.9%
Nickel, wire reel, 100m, diameter 0.25mm, hard, 99.98%
Nickel, foil, 0.5m coil, thickness 0.15mm, annealed, 99%
Nickel, foil, 4mm disks, thickness 0.009mm, 99.95%
Nickel, foil, 0.5m coil, thickness 0.0075mm, 99.9%
Nickel, foil, 0.5m coil, thickness 0.0125mm, annealed, 99%
Nickel, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.99+%
Nickel, pellets, 50g, max. size 5mm, 99.99+%
Nickel, wire reel, 10m, diameter 0.125mm, hard, 99.98%
Nickel, rod, 1000mm, diameter 6.35mm, 99%
Nickel, foil, 0.5m coil, thickness 1.0mm, annealed, 99%