Merck
  • Aluminum disruption of calcium homeostasis and signal transduction resembles change that occurs in aging and Alzheimer's disease.
Aluminum disruption of calcium homeostasis and signal transduction resembles change that occurs in aging and Alzheimer's disease.
Journal of Alzheimer's disease : JAD (2012-02-15)
J R Walton
ABSTRACT
Most humans living in industrialized societies are routinely exposed to bioavailable aluminum salts in the form of additives-in commercially-prepared foods, alum-clarified drinking water, certain pharmaceuticals, sunscreens, and other topical applications. Minute amounts of this aluminum are absorbed into the circulation. Trace aluminum levels cross the blood-brain barrier and progressively accumulate in large pyramidal neurons of the hippocampus, cortex, and other brain regions vulnerable in Alzheimer's disease. More aluminum enters the brain than leaves, resulting in a net increase in intraneuronal aluminum with advancing age. Aluminum is responsible for two main types of toxic damage in cells. As a pro-oxidant, aluminum causes oxidative damage both on its own and in synergy with iron. Aluminum also competes with, and substitutes for, essential metals-primarily Mg2+, iron and Ca2+ ions-in or on proteins and their co-factors. The author hypothesizes that intraneuronal aluminum interferes with Ca2+ metabolism in the aged brain and describes a way to test this hypothesis. This paper reviews: 1) major changes that occur in brain Ca2+ homeostasis and Ca2+ signaling, subtly with aging and more overtly in Alzheimer's disease; and 2) evidence from the scientific literature that aluminum causes these same changes in neurons.
Sigma-Aldrich
Aluminum, powder, <5 μm particle size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Aluminum, powder, <75 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, ≤30 μm, spherical powder, ≥99%
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Aluminum, insulated wire, 20m, conductor diameter 0.1mm, insulation thickness 0.005mm, polyimide insulation, 99.95+%
Aluminum, mesh, 900x900mm, nominal aperture 0.38mm, wire diameter 0.25mm, 40x40 wires/inch, open area 37%, plain weave mesh
Aluminum, microfoil, disks, 25mm, thinness 1.0μm, specific density 269.9μg/cm2, permanent mylar 3.5μm support, 99.999%
Aluminum, insulated wire, 2m, conductor diameter 0.1mm, insulation thickness 0.005mm, polyimide insulation, 99.95+%
Aluminum, microleaf disks, 25mm, thinness 0.15μm, specific density 41.28μg/cm2, removable support, 99.999%
Aluminum, microleaf disks, 25mm, thinness 0.25μm, specific density 68.8μg/cm2, removable support, 99.999%
Aluminum, sphere, 100pcs, diameter 6.0mm, 99%
Aluminum, tube, 500mm, outside diameter 3.0mm, inside diameter 1.62mm, wall thickness 0.69mm, hard, 99.5%
Aluminum, microleaf disks, 25mm, thinness 0.5μm, specific density 120.6μg/cm2, removable support, 99.999%
Aluminum, rod, 100mm, diameter 28mm, as drawn, 99.999%
Aluminum, wire reel, 500m, diameter 1.0mm, as drawn, 99.95+%
Aluminum, wire reel, 20m, diameter 0.02mm, hard, 99.99%
Aluminum, wire reel, 10m, diameter 1.5mm, as drawn, 99.999%
Aluminum, wire reel, 500m, diameter 0.30mm, as drawn, 99.5%
Aluminum, wire reel, 10m, diameter 0.015mm, hard, 99.99%
Aluminum, wire reel, 2m, diameter 0.25mm, as drawn, 99.999%
Aluminum, wire reel, 200m, diameter 0.25mm, as drawn, 99.95+%
Aluminum, wire reel, 20m, diameter 0.25mm, hard, 99.99+%