Merck
Hepatic response to aluminum toxicity: dyslipidemia and liver diseases.
Experimental cell research (2011-07-27)
Ryan J Mailloux, Joseph Lemire, Vasu D Appanna
ABSTRACT
Aluminum (Al) is a metal toxin that has been implicated in the etiology of a number of diseases including Alzheimer's, Parkinson's, dialysis encephalopathy, and osteomalacia. Al has been shown to exert its effects by disrupting lipid membrane fluidity, perturbing iron (Fe), magnesium, and calcium homeostasis, and causing oxidative stress. However, the exact molecular targets of aluminum's toxicity have remained elusive. In the present review, we describe how the use of a systems biology approach in cultured hepatoblastoma cells (HepG2) allowed the identification of the molecular targets of Al toxicity. Mitochondrial metabolism is the main site of the toxicological action of Al. Fe-dependent and redox sensitive enzymes in the tricarboxylic acid (TCA) cycle and oxidative phosphorylation (OXPHOS) are dramatically decreased by Al exposure. In an effort to compensate for diminished mitochondrial function, Al-treated cells stabilize hypoxia inducible factor-1α (HIF-1α) to increase ATP production by glycolysis. Additionally, Al toxicity leads to an increase in intracellular lipid accumulation due to enhanced lipogenesis and a decrease in the β-oxidation of fatty acids. Central to these effects is the alteration of α-ketoglutarate (KG) homeostasis. In Al-exposed cells, KG is preferentially used to quench ROS leading to succinate accumulation and HIF-1α stabilization. Moreover, the channeling of KG to combat oxidative stress leads to a reduction of l-carnitine biosynthesis and a concomitant decrease in fatty acid oxidation. The fluidity and interaction of these metabolic modules and the implications of these findings in liver-related disorders are discussed herein.
Sigma-Aldrich
Aluminum, powder, <5 μm particle size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Aluminum, powder, <75 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, ≤30 μm, spherical powder, ≥99%
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Aluminum, insulated wire, 20m, conductor diameter 0.1mm, insulation thickness 0.005mm, polyimide insulation, 99.95+%
Aluminum, mesh, 900x900mm, nominal aperture 0.38mm, wire diameter 0.25mm, 40x40 wires/inch, open area 37%, plain weave mesh
Aluminum, microfoil, disks, 25mm, thinness 1.0μm, specific density 269.9μg/cm2, permanent mylar 3.5μm support, 99.999%
Aluminum, insulated wire, 2m, conductor diameter 0.1mm, insulation thickness 0.005mm, polyimide insulation, 99.95+%
Aluminum, microleaf disks, 25mm, thinness 0.15μm, specific density 41.28μg/cm2, removable support, 99.999%
Aluminum, microleaf disks, 25mm, thinness 0.25μm, specific density 68.8μg/cm2, removable support, 99.999%
Aluminum, sphere, 100pcs, diameter 6.0mm, 99%
Aluminum, tube, 500mm, outside diameter 3.0mm, inside diameter 1.62mm, wall thickness 0.69mm, hard, 99.5%
Aluminum, microleaf disks, 25mm, thinness 0.5μm, specific density 120.6μg/cm2, removable support, 99.999%
Aluminum, rod, 100mm, diameter 28mm, as drawn, 99.999%
Aluminum, wire reel, 500m, diameter 1.0mm, as drawn, 99.95+%
Aluminum, wire reel, 20m, diameter 0.02mm, hard, 99.99%
Aluminum, wire reel, 10m, diameter 1.5mm, as drawn, 99.999%
Aluminum, wire reel, 500m, diameter 0.30mm, as drawn, 99.5%
Aluminum, wire reel, 10m, diameter 0.015mm, hard, 99.99%
Aluminum, wire reel, 2m, diameter 0.25mm, as drawn, 99.999%
Aluminum, wire reel, 200m, diameter 0.25mm, as drawn, 99.95+%
Aluminum, wire reel, 20m, diameter 0.25mm, hard, 99.99+%