Merck
Diethylene glycol poisoning.
Clinical toxicology (Philadelphia, Pa.) (2009-07-10)
Leo J Schep, Robin J Slaughter, Wayne A Temple, D Michael G Beasley
ABSTRACT
Diethylene glycol (DEG) is a clear, colorless, practically odorless, viscous, hygroscopic liquid with a sweetish taste. In addition to its use in a wide range of industrial products, it has also been involved in a number of prominent mass poisonings spanning back to 1937. Despite DEG's toxicity and associated epidemics of fatal poisonings, a comprehensive review has not been published. A summary of the literature on DEG was compiled by systematically searching OVID MEDLINE and ISI Web of Science. Further information was obtained from book chapters, relevant news reports, and web material. The aim of this review is to summarize all main aspects of DEG poisoning including epidemiology, toxicokinetics, mechanisms of toxicity, clinical features, toxicity of DEG, diagnosis, and management. Most of the documented cases of DEG poisoning have been epidemics (numbering over a dozen) where DEG was substituted in pharmaceutical preparations. More often, these epidemics have occurred in developing and impoverished nations where there is limited access to intensive medical care and quality control procedures are substandard. Following ingestion, DEG is rapidly absorbed and distributed within the body, predominantly to regions that are well perfused. Metabolism occurs principally in the liver and both the parent and the metabolite, 2-hydroxyethoxyacetic acid (HEAA), are renally eliminated rapidly. Although the mechanism of toxicity is not clearly elucidated, research suggests that the DEG metabolite, HEAA, is the major contributor to renal and neurological toxicities. The clinical effects of DEG poisoning can be divided into three stages: The first phase consists of gastrointestinal symptoms with evidence of inebriation and developing metabolic acidosis. If poisoning is pronounced, patients can progress to a second phase with more severe metabolic acidosis and evidence of emerging renal injury, which, in the absence of appropriate supportive care, can lead to death. If patients are stabilized, they may then enter the final phase with various delayed neuropathies and other neurological effects, sometimes fatal. TOXICITY OF DEG: Doses of DEG necessary to cause human morbidity and mortality are not well established. They are based predominantly on reports following some epidemics of mass poisonings, which may underestimate toxicity. The mean estimated fatal dose in an adult has been defined as approximately 1 mL/kg of pure DEG. Initial treatment consists of appropriate airway management and attention to acid-base abnormalities. Prompt use of fomepizole or ethanol is important in preventing the formation of the toxic metabolite HEAA; hemodialysis can also be critical, and assisted ventilation may be required. DEG ingestion can lead to serious complications that may prove fatal. Prognosis may be improved, however, with prompt supportive care and timely use of fomepizole or ethanol.
Sigma-Aldrich
Diethylene glycol, ReagentPlus®, 99%
Sigma-Aldrich
Diethylene glycol, puriss. p.a., ≥99.0% (GC), colorless
Sigma-Aldrich
Diethylene glycol, BioUltra, ≥99.0% (GC)
Supelco
Diethylene glycol, analytical standard
Supelco
Diethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diethylene glycol, SAJ first grade, ≥98.0%
USP
Diethylene glycol, United States Pharmacopeia (USP) Reference Standard