Merck
  • Trimethoprim and enterococci in urinary tract infections: new perspectives on an old issue.
Trimethoprim and enterococci in urinary tract infections: new perspectives on an old issue.
The Journal of antimicrobial chemotherapy (2008-04-15)
Karin Tegmark Wisell, Gunnar Kahlmeter, Christian G Giske
ABSTRACT
The lack of oral treatment alternatives for enterococcal urinary tract infections (UTIs) has led to a renewed interest in trimethoprim. Enterococci can incorporate exogenously produced folates and thereby reverse the effect of trimethoprim. Although a large proportion of enterococci appear susceptible to trimethoprim in vitro using standard media devoid of folates, a 360-fold increase in the MIC can be seen when susceptibility testing is performed in media containing fresh urine. Even if trimethoprim has a favourable pharmacokinetic profile, with high serum and very high urine concentrations, pharmacodynamic (PD) estimates show that a large proportion of the apparent wild-type isolates (as categorized by standard susceptibility testing) have unfavourable PD indices. The clinical efficacy of trimethoprim in enterococcal UTI is debated. We could identify not more than 38 evaluable cases of enterococcal UTI in the literature. The eradication rate was 82%. Case reports where patients on co-trimoxazole for UTI have developed bacteraemia with enterococci susceptible to trimethoprim seem to support experimental findings that standard antimicrobial susceptibility testing poorly predicts the clinical outcome of trimethoprim therapy. The European Committee on Antimicrobial Susceptibility Testing and the national breakpoint committees in Europe have recently debated the role of trimethoprim in the treatment of enterococcal UTI and agreed to categorize wild-type enterococci as intermediate to trimethoprim and trimethoprim/sulfamethoxazole. This allows the distinction between enterococci with and without acquired resistance mechanisms to trimethoprim. This review discusses the microbiological, experimental, clinical and PD aspects of the usage of trimethoprim for enterococcal UTI.
Product Description
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Trimethoprim, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Trimethoprim, ≥98.5%
Trimethoprim, European Pharmacopoeia (EP) Reference Standard