Sigma-Aldrich
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, 60-100 mesh, 99.9%
Tantalum, foil, 10mm disks, thickness 0.0125mm, 99.9%
Tantalum, foil, 10mm disks, thickness 0.0075mm, 99.9%
Tantalum, foil, 10mm disks, thickness 0.015mm, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.01mm, 99.9%
Tantalum, foil, 6mm disks, thickness 0.015mm, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.0015mm, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.01mm, 99.9%
Tantalum, rod, 200mm, diameter 1.5mm, annealed, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.001mm, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.0075mm, 99.9%
Tantalum, rod, 100mm, diameter 10.0mm, annealed, 99.9%
Tantalum, foil, not light tested, 100x100mm, thickness 0.009mm, 99.9%
Tantalum, tube, 100mm, outside diameter 7.0mm, inside diameter 6.7mm, wall thickness 0.15mm, as drawn, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.0015mm, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.034mm, as rolled, 99.9%
Tantalum, mesh, 40x40mm, nominal aperture 0.43mm, wire diameter 0.075mm, 50x50 wires/inch, open area 72%, plain weave mesh
Tantalum, foil, not light tested, 100x100mm, thickness 0.003mm, 99.9%
Tantalum, rod, 50mm, diameter 18mm, annealed, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.015mm, 99.9%
Tantalum, tube, 100mm, outside diameter 12.7mm, inside diameter 11.7mm, wall thickness 0.5mm, annealed, 99.9%
Tantalum, tube, 1000mm, outside diameter 2.0mm, inside diameter 1.4mm, wall thickness 0.3mm, as drawn, 99.9%
Tantalum, rod, 500mm, diameter 1.0mm, 99.9%
Tantalum, foil, light tested, 100x100mm, thickness 0.034mm, as rolled, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.01mm, 99.9%
Tantalum, rod, 1000mm, diameter 1.0mm, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.003mm, 99.9%
Tantalum, rod, 500mm, diameter 1.5mm, annealed, 99.9%
Tantalum, tube, 100mm, outside diameter 8.7mm, inside diameter 8.34mm, wall thickness 0.18mm, as drawn, 99.9%