Merck
  • CD79a: a novel marker for B-cell neoplasms in routinely processed tissue samples.
CD79a: a novel marker for B-cell neoplasms in routinely processed tissue samples.
Blood (1995-08-15)
D Y Mason, J L Cordell, M H Brown, J Borst, M Jones, K Pulford, E Jaffe, E Ralfkiaer, F Dallenbach, H Stein
ABSTRACT
The CD79 molecule, comprising two polypeptide chains, mb-1 (CD79a) and B29 (CD79b), is physically associated in the B-cell membrane with immunoglobulin. It transmits a signal after antigen binding and may, therefore, be considered the B cell equivalent of CD3. It appears before the pre-B-cell stage, and the mb-1 (CD79a) chain can still be present at the plasma cell stage. In this report, we describe a new anti-CD79a monoclonal antibody, JCB117, which reacts with human B cells in paraffin embedded tissue sections, including decalcified bone marrow trephines. When tested on a total of 454 paraffin embedded tissue biopsies, gathered from a number of different institutions, it reacted with the great majority (97%) of B-cell neoplasms, covering the full range of B-cell maturation, including 10 of 20 cases of myeloma/plasmacytoma. It is of interest that the antibody labels precursor B-cell acute lymphoblastic leukemia samples, making it the most reliable B-cell marker detectable in paraffin-embedded specimens in this disorder. All neoplasms of T cell or nonlymphoid origin were negative, indicating that antibody JCB117 may be of value to diagnostic histopathologists for the identification of B-cell neoplasms of all maturation stages.
Sigma-Aldrich
CD79a (JCB117) Mouse Monoclonal Antibody
Sigma-Aldrich
CD79a (SP18) Rabbit Monoclonal Antibody