Merck
  • Extreme masking: achieving predictable outcomes in challenging situations with lithium disilicate bonded restorations.
Extreme masking: achieving predictable outcomes in challenging situations with lithium disilicate bonded restorations.
The international journal of esthetic dentistry (2014-04-26)
Yugo Hatai
ABSTRACT
In contemporary dentistry, we have a vast range of materials to choose from, and metal free restorations have become the premier materials for achieving the ultimate in both esthetics and durability. Metal-free restorations are utilized with more conservative preparations to preserve the vital natural dentition, and have proven to be superior alternatives to traditional porcelain-fused-to-metal (PFM) restorations in many cases. There are always "pros and cons" when selecting materials, and to make the best choice it is essential for dental professionals to plan precisely and understand their options in any clinical situation. Selecting suitable materials and techniques involves consideration of the following factors: - Esthetic zone. - Required strength based on the patient's occlusion/dental habits. - Preparation reduction. - Position of the margin. - Type of restoration/preparation. - The treating clinician's philosophy. - Stump shade. Final shade. One of the most significant challenges in the metal-free dentistry is the reproduction of natural dentition without the influence of a "negative stump" - a very dark or metal core showing through the final restorations. There are many factors to be considered when working on such a case, and controlling the opacity of the coping and crown is the key to success. This article presents a unique "outside of the box" technique that provides consistent, predictable and durable restorations, which provide the best possible esthetic outcome.
Sigma-Aldrich
Zirconium(IV) oxide, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Zirconium(IV) oxide, powder, 5 μm, 99% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, 99.99% trace metals basis (purity excludes ~2% HfO2)
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Zirconium, foil, 100x100mm, thickness 2.0mm, annealed, 99.2%
Zirconium, foil, 100x100mm, thickness 0.125mm, annealed, 99.2%
Zirconium, foil, 25x25mm, thickness 0.125mm, annealed, 99.2%
Zirconium, wire reel, 5m, diameter 0.05mm, as drawn, 99.2%
Zirconium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.2%
Zirconium, wire reel, 1m, diameter 0.28mm, annealed, 99.2%
Zirconium, tube, 200mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, annealed, 99.2%
Zirconium, rod, 500mm, diameter 5.0mm, centerless ground, 99.2%
Zirconium, rod, 200mm, diameter 2.0mm, centerless ground, 99.2%
Zirconium, tube, 1000mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, annealed, 99.2%
Zirconium, tube, 200mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, annealed, 99.2%
Zirconium, sponge, 20g, max. size 25mm, 99.2%
Zirconium, rod, 100mm, diameter 9.5mm, centerless ground, 99.2%
Zirconium, wire reel, 5m, diameter 0.25mm, as drawn, 99.2%
Zirconium, tube, 1000mm, outside diameter 6.35mm, inside diameter 4.55mm, wall thickness 0.9mm, annealed, 99.2%
Zirconium, rod, 200mm, diameter 3.0mm, centerless ground, 99.2%
Zirconium, tube, 100mm, outside diameter 6.35mm, inside diameter 4.55mm, wall thickness 0.9mm, annealed, 99.2%
Zirconium, rod, 100mm, diameter 6.0mm, centerless ground, 99.2%
Zirconium, rod, 500mm, diameter 12.7mm, centerless ground, 99.2%
Zirconium, wire reel, 5m, diameter 0.28mm, annealed, 99.2%
Zirconium, wire reel, 10m, diameter 0.25mm, as drawn, 99.2%