Merck
  • Effects of sub-chronic aluminum chloride on spermatogenesis and testicular enzymatic activity in male rats.
Effects of sub-chronic aluminum chloride on spermatogenesis and testicular enzymatic activity in male rats.
Life sciences (2014-03-13)
Y Z Zhu, H Sun, Yang Fu, J Wang, M Song, M Li, Y F Li, L G Miao
ABSTRACT
The aim of this study was to determine the effects of sub-chronic aluminum chloride (AlCl3) on spermatogenesis and testicular enzymatic activity in male rats. Forty Wistar male rats were randomly divided into four groups: control group (CG, 0), low-dose group (LG, 64.18 mg/kg BW AlCl3), mid-dose group (MG, 128.36 mg/kg BW AlCl3) and high-dose group (HG, 256.72 mg/kg BW AlCl3). The rats were orally administered with AlCl3 for 120 days. At the end of the experiment, the contents of Al, Fe, Cu and Zn, the enzyme activities of testicular acid phosphatase (ACP), succinate dehydrogenase (SDH), lactate dehydrogenase (LDH), lactate dehydrogenase isoenzyme (LDH-x), the sperm count and the sperm malformation rate were examined. The results showed that the Al and Cu contents, sperm count and the enzyme activities of testicular ACP, SDH, LDH and LDH-x decreased, while the Zn and Fe contents and sperm malformation rate increased in AlCl3-treated rats. It suggests that sub-chronic AlCl3 disorders the balance of trace element and decreases the spermatogenesis and the activities of testicular enzymes, indicating that AlCl3 has adverse effect on the testicular function in male rats.
Sigma-Aldrich
Aluminum chloride, SAJ first grade, ≥98.0%
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Zinc, wire reel, 25m, diameter 0.125mm, as drawn, 99%
Zinc, wire reel, 2m, diameter 0.5mm, as drawn, 99.99+%
Zinc, wire reel, 10m, diameter 1.0mm, 99.99+%
Zinc, wire reel, 10m, diameter 2.0mm, 99.99+%
Sigma-Aldrich
Aluminum chloride, reagent grade, 98%
Sigma-Aldrich
Aluminum chloride solution, 1.0 M in nitrobenzene
Sigma-Aldrich
Aluminum chloride, anhydrous, sublimed, ≥98%
Sigma-Aldrich
Aluminum chloride, ReagentPlus®, 99%
Zinc, foil, 15mm disks, thickness 0.05mm, as rolled, 99.99+%
Zinc, foil, 10mm disks, thickness 0.0025mm, 99.9%
Zinc, foil, 0.5m coil, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.35mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 2.0mm, as rolled, 99.95+%
Zinc, foil, 0.3m coil, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.25mm, as rolled, 99.95+%