- sp3-sp3 Coupling reactions in the synthesis of natural products and biologically active molecules.
sp3-sp3 Coupling reactions in the synthesis of natural products and biologically active molecules.
Natural product reports (2014-02-28)
Egor Geist, Andreas Kirschning, Thomas Schmidt
PMID24573302
ABSTRACT
This Highlight covers the current status of relatively unexplored sp(3)-sp(3) cross-coupling reactions with particular focus on natural product and related syntheses.
Product Description
Nickel, rod, 200mm, diameter 12.7mm, 99.98%
Nickel, foil, 0.5m coil, thickness 0.05mm, 99.9%
Nickel, wire reel, 10m, diameter 0.025mm, as drawn, 99.98%
Nickel, foil, 50x50mm, thickness 0.3mm, annealed, 99%
Nickel, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Nickel, foil, 0.5m coil, thickness 0.15mm, annealed, 99%
Nickel, wire reel, 100m, diameter 0.5mm, as drawn, 99%
Nickel, wire reel, 0.5m, diameter 0.05mm, as drawn, 99.99+%
Nickel, foil, 2m coil, thickness 0.25mm, annealed, 99.98%
Nickel, foil, 25x25mm, thickness 3.2mm, as rolled, 99.99+%
Nickel, foil, 50x50mm, thickness 0.125mm, as rolled, 99.999%
Nickel, foil, 25x25mm, thickness 0.3mm, annealed, 99%
Nickel, foil, 0.5m coil, thickness 1.0mm, annealed, 99%
Nickel, foil, 4mm disks, thickness 0.025mm, 99.9%
Nickel, foil, 0.3m coil, thickness 0.125mm, as rolled, 99.99+%
Nickel, foil, 50x50mm, thickness 1.0mm, as rolled, 99.99+%
Nickel, tube, 500mm, outside diameter 1.0mm, inside diameter 0.8mm, wall thickness 0.1mm, hard, 99.5%
Nickel, pellets, 20g, max. size 20mm, 99.99+%
Nickel, foil, 10m coil, thickness 0.125mm, coil width 50mm, annealed, 99%
Nickel, rod, 100mm, diameter 10.0mm, 99.99+%
Nickel, foil, 0.5m coil, thickness 0.005mm, as rolled, 99.9%
Nickel, foil, 50x50mm, thickness 0.3mm, as rolled, 99.999%
Nickel, foil, 10mm disks, thickness 0.025mm, as rolled, 99.99+%
Nickel, foil, 25x25mm, thickness 2.0mm, annealed, 99%
Nickel, foil, 50x50mm, thickness 1.0mm, annealed, 99%
Nickel, foil, 10mm disks, thickness 0.05mm, annealed, 99%
Nickel, tube, 500mm, outside diameter 3.25mm, inside diameter 3.05mm, wall thickness 0.1mm, hard, 99.5%
Nickel, foil, 10mm disks, thickness 0.005mm, as rolled, 99.9%
Nickel, rod, 1000mm, diameter 2.0mm, 99.99+%
Nickel, foil, 2m coil, thickness 0.5mm, annealed, 99.98%