Merck
  • Dietary acrylamide intake and the risk of colorectal cancer with specific mutations in KRAS and APC.
Dietary acrylamide intake and the risk of colorectal cancer with specific mutations in KRAS and APC.
Carcinogenesis (2014-01-09)
Janneke G F Hogervorst, Daisy de Bruijn-Geraets, Leo J Schouten, Manon van Engeland, Theo M C M de Kok, R Alexandra Goldbohm, Piet A van den Brandt, Matty P Weijenberg
ABSTRACT
Acrylamide, a probable human carcinogen, is present in heat-treated carbohydrate-rich foods. Epidemiological studies have not shown a clear association between acrylamide intake and colorectal cancer (CRC) risk. This may be due to the molecular heterogeneity in colorectal tumors, which was not taken into consideration before. Since the acrylamide metabolite glycidamide induces specific DNA mutations in rodents, we investigated whether acrylamide is associated with CRC risk characterized by mutations in Kirsten-ras (KRAS) and adenomatous polyposis coli (APC); key genes in colorectal carcinogenesis. This case-cohort analysis, within the Netherlands Cohort Study on diet and cancer, was based on 7.3 years of follow-up. Acrylamide intake was assessed with a food frequency questionnaire. Mutation analysis of codons 1286-1520 in exon 15 in APC and codons 12 and 13 in exon 1 in KRAS was performed on tumor tissue of 733 cases. Hazard ratios (HR) were calculated using Cox proportional hazards analysis. Among men, acrylamide intake was statistically significantly associated with an increased risk of particularly tumors with an activating KRAS mutation {HR fourth versus first quartile: 2.12 [95% confidence interval (CI): 1.16-3.87], P trend: 0.01}. Among women, acrylamide intake was statistically significantly associated with a decreased risk of particularly tumors with a truncating APC mutation (fourth versus first quartile: 0.47 (95% CI: 0.23-0.94), P trend: 0.02), but only in the highest quartile of intake. This is the first study to show that acrylamide might be associated with CRC with specific somatic mutations, differentially in men and women. More research is needed to corroborate or refute these findings.
Sigma-Aldrich
Acrylamide, SAJ first grade, ≥98.0%
Sigma-Aldrich
Acrylamide, ≥99.9%
Sigma-Aldrich
Acrylamide, ≥98.0%
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Supelco
Acrylamide solution, 40% in H2O, for molecular biology
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered