Merck
  • Peg-IFNα/ribavirin/protease inhibitor combination in hepatitis C virus associated mixed cryoglobulinemia vasculitis: results at week 24.
Peg-IFNα/ribavirin/protease inhibitor combination in hepatitis C virus associated mixed cryoglobulinemia vasculitis: results at week 24.
Annals of the rheumatic diseases (2013-04-23)
David Saadoun, M Resche Rigon, V Thibault, M Longuet, S Pol, F Blanc, G Pialoux, A Karras, D Bazin-Karra, C Cazorla, D Vittecoq, L Musset, O Decaux, J M Ziza, O Lambotte, Patrice Cacoub
ABSTRACT
The standard-of-care treatment of patients with hepatitis C virus (HCV)-mixed cryoglobulinemia (MC) vasculitis includes pegylated interferon α (PegIFN)-α plus ribavirin and/or rituximab. About 30-40% of patients are non-responders or relapsers to such combination. To analyse the safety and efficacy of Peg-IFNα/ribavirin/protease inhibitor combination in HCV-MC vasculitis. Open-label, prospective, cohort study including 23 patients with HCV-MC vasculitis. Peg-IFNα/ribavirin was associated to telaprevir (375 mg three times daily, for 12 weeks, (n=15)) or boceprevir (800 mg three times daily, for 44 weeks, (n=8)) for 48 weeks. The median age was 59 (52.5-66) years, with 48.8% women. Thirteen patients (56.5%) were complete clinical responders, and 10 (43.5%) were partial responders at week 24. The virological response (ie, HCV RNA negativation) was of 69.6% at week 24 (p=0.005). The cryoglobulin level decreased from 0.44 to 0.06 g/l (p=0.0006) and the C4 level increased from 0.09 to 0.15 g/l (p=0.045). Grades 3 and 4 adverse events (mainly anaemia, neutropenia and thrombocytopenia) were observed in 10 cases (43.5%). Twenty patients (87%) received erythropoietin, 9 (39.1%) had red cell transfusion, and 2 (8.7%) had granulocyte stimulating agents. Antiviral therapy discontinuation was required in 8 (34.7%) patients for virological non-response (n=5), virological relapse (n=2) and depression (n=1). Peg-IFNα/ribavirin/protease inhibitor combination seems highly effective in HCV-MC. Such therapeutic regimen should be administered cautiously considering the high rate of side effects.
Product Description
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Proline, SAJ special grade, ≥99.0%
USP
L-Proline, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Proline
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Proline, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)