Merck
  • Direct reversal of glucocorticoid resistance by AKT inhibition in acute lymphoblastic leukemia.
Direct reversal of glucocorticoid resistance by AKT inhibition in acute lymphoblastic leukemia.
Cancer cell (2013-12-03)
Erich Piovan, Jiyang Yu, Valeria Tosello, Daniel Herranz, Alberto Ambesi-Impiombato, Ana Carolina Da Silva, Marta Sanchez-Martin, Arianne Perez-Garcia, Isaura Rigo, Mireia Castillo, Stefano Indraccolo, Justin R Cross, Elisa de Stanchina, Elisabeth Paietta, Janis Racevskis, Jacob M Rowe, Martin S Tallman, Giuseppe Basso, Jules P Meijerink, Carlos Cordon-Cardo, Andrea Califano, Adolfo A Ferrando
ABSTRACT
Glucocorticoid resistance is a major driver of therapeutic failure in T cell acute lymphoblastic leukemia (T-ALL). Here, we identify the AKT1 kinase as a major negative regulator of the NR3C1 glucocorticoid receptor protein activity driving glucocorticoid resistance in T-ALL. Mechanistically, AKT1 impairs glucocorticoid-induced gene expression by direct phosphorylation of NR3C1 at position S134 and blocking glucocorticoid-induced NR3C1 translocation to the nucleus. Moreover, we demonstrate that loss of PTEN and consequent AKT1 activation can effectively block glucocorticoid-induced apoptosis and induce resistance to glucocorticoid therapy. Conversely, pharmacologic inhibition of AKT with MK2206 effectively restores glucocorticoid-induced NR3C1 translocation to the nucleus, increases the response of T-ALL cells to glucocorticoid therapy, and effectively reverses glucocorticoid resistance in vitro and in vivo.
Product Description
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®