Merck
  • Isoflurane suppresses cortical spreading depolarizations compared to propofol--implications for sedation of neurocritical care patients.
Isoflurane suppresses cortical spreading depolarizations compared to propofol--implications for sedation of neurocritical care patients.
Experimental neurology (2013-11-20)
Masatoshi Takagaki, Delphine Feuerstein, Tetsuya Kumagai, Markus Gramer, Toshiki Yoshimine, Rudolf Graf
ABSTRACT
Sedatives in the neurointensive care unit can strongly influence patients' risks of developing secondary brain damage. In particular, isoflurane, a volatile anesthetic, has been recently re-introduced to the neurointensive care unit, and first clinical studies suggest beneficial effects due to elevation of cerebral blood flow and reduction of metabolism. In contrast, propofol is a commonly used intravenous sedative that reduces cerebral blood flow and intra-cranial pressure. We have here studied the influence of these two sedatives on the occurrence of cortical spreading depolarizations (CSDs), which have emerged over the last decade as a major mechanism of delayed brain injury in stroke and brain trauma, constituting a substantial vascular and metabolic threat to peri-infarct tissue and being associated with poor patient outcome. Two experimental models were tested in Wistar rats anesthetized either with isoflurane or with propofol: KCl-evoked CSDs (n=10) and spontaneous CSDs after occlusion of the middle cerebral artery (n=14). Spatiotemporal patterns of CSD waves were observed by real-time laser speckle imaging of regional cerebral blood flow changes associated with the CSDs. During 30 min of cortical KCl application, 5.2±0.7 CSDs were induced under isoflurane compared to 10.2±1.8 CSDs under propofol (p<0.001). After focal ischemia, 2.43±1.0 CSDs/h emerged spontaneously under isoflurane versus 6.83±2.5 CSDs/h under propofol (p<0.001). Furthermore, baseline blood flow and glycemia were much higher under isoflurane compared to propofol, which may set the tissue in better metabolic conditions to recover from the occurrence of CSD waves. We conclude that isoflurane, in comparison to propofol, decreases the occurrence of CSDs and may improve recovery from these metabolically demanding waves. To reduce CSD induced secondary tissue damage, we suggest isoflurane to be favored over propofol to sedate acute stroke and trauma patients in the neurointensive care unit.
Product Description
Supelco
Propofol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Propofol for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, ~3 M KCl, saturated with silver chloride
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride solution, 0.01 M
Sigma-Aldrich
Potassium standard solution, suitable for atomic absorption spectrometry, 1000 ppm K+, 1 mg/mL K+
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Propofol, European Pharmacopoeia (EP) Reference Standard
Isoflurane, European Pharmacopoeia (EP) Reference Standard