Merck
  • Essential oil from Chenopodium ambrosioides and main components: activity against Leishmania, their mitochondria and other microorganisms.
Essential oil from Chenopodium ambrosioides and main components: activity against Leishmania, their mitochondria and other microorganisms.
Experimental parasitology (2013-11-05)
Lianet Monzote, Marley García, Jacinta Pastor, Lizette Gil, Ramón Scull, Louis Maes, Paul Cos, Lars Gille
ABSTRACT
Chenopodium ambrosioides is an aromatic herb used by native people to treat parasitic diseases. The aim of this work is to compare the in vitro anti-leishmanial activity of the essential oil (EO) from C. ambrosioides and its major components (ascaridole, carvacrol and caryophyllene oxide) and study their mechanism of action and activity against a panel of microorganism. Antileishmanial activity and cytotoxicity of the EO and major components was study. In addition, experiments to elucidate the mechanism of action were perform and activities against other microorganisms (bacteria, fungi and protozoa) were evaluate. All products were active against promastigote and amastigote forms of Leishmania. Ascaridole exhibited the better antileishmanial activity and the EO the highest selectivity index. The exploration of the mechanism suggests that the products cause a breakdown of mitochondrial membrane potential and a modification of redox indexes. Only EO showed antiprotozoal effect against Plasmodium falciparum and Trypanosoma brucei; while no activity against bacteria and fungi was observed. Our results demonstrate the potentialities of EO in cellular and molecular system, which could be consider in future studies to develop new antileishmanial drugs with a wide anti-parasitic spectrum.
Sigma-Aldrich
Cytochrome c from equine heart, BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis
Sigma-Aldrich
Cytochrome c from pigeon breast muscle, ≥95% based on Mol. Wt. 12,173 basis
Sigma-Aldrich
Cytochrome c from equine heart, BioReagent, suitable for GFC marker
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% (SDS-PAGE)
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% based on Mol. Wt. 12,384 basis
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture
Sigma-Aldrich
ProteoMass Cytochrome c MALDI-MS Standard, vial of 10 nmol, (M+H+) 12,361.96 Da by calculation
Sigma-Aldrich
Cytochrome c from Saccharomyces cerevisiae, ≥85% based on Mol. Wt. 12,588 basis