Merck
  • Performance enhancement of metal-oxide-semiconductor tunneling temperature sensors with nanoscale oxides by employing ultrathin Al2O3 high-k dielectrics.
Performance enhancement of metal-oxide-semiconductor tunneling temperature sensors with nanoscale oxides by employing ultrathin Al2O3 high-k dielectrics.
Nanoscale (2013-07-25)
Chien-Chih Lin, Jenn-Gwo Hwu
ABSTRACT
We demonstrated a promising route for enhancing temperature sensitivity, improving saturation voltage, and reducing power consumption of the MOS(p) tunneling temperature sensors by introducing ultrathin Al2O3 into the dielectric stacks. Detailed illustrations of the working mechanism and device concept are given in this work. Three kinds of MOS(p) tunneling temperature sensors with nanoscale SiO2, HfO2, and Al2O3 dielectrics were compared comprehensively. For Al2O3 MOS(p) devices with an equivalent oxide thickness of 2 nm, the sensing performance was effectively improved and the temperature-sensitive current-voltage characteristics are reliable and reproducible. The low-temperature processing Al2O3 MOS(p) tunneling temperature sensors are potential candidates for temperature monitoring sensors on chips or biomedical systems under low thermal budget processing consideration.
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, CP
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, Brockmann I, standard grade, activated, basic
Sigma-Aldrich
Aluminum oxide, Brockmann I, standard grade, activated, acidic
Sigma-Aldrich
Aluminum oxide, activated, Brockmann I, standard grade, neutral
Sigma-Aldrich
Aluminum oxide, Brockmann I, activated, standard grade, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, standard grade, activated, Brockmann I, neutral, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, pellets, 3 mm
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I