Merck
  • Treatment of onychomycosis using a submillisecond 1064-nm neodymium:yttrium-aluminum-garnet laser.
Treatment of onychomycosis using a submillisecond 1064-nm neodymium:yttrium-aluminum-garnet laser.
Journal of the American Academy of Dermatology (2013-07-17)
Caitlin Carney, Wendy Cantrell, Judy Warner, Boni Elewski
ABSTRACT
Laser treatment has emerged as a novel treatment modality for onychomycosis. We sought to determine thermal response and optical effects of a submillisecond neodymium:yttrium-aluminum-garnet (Nd:YAG) 1064-nm laser on common fungal nail pathogens, and the clinical efficacy and safety of the Nd:YAG 1064-nm laser on onychomycotic toenails. A 4-part in vitro and in vivo study was conducted using a Nd:YAG 1064-nm laser. The first portion evaluated 3 different nail pathogens in suspension at 7 heat and time exposures. The second and third parts of the study irradiated pure fungal colonies. The final portion involved an in vivo treatment of toenails over 5 treatment sessions. A fungicidal effect for Trichophyton rubrum was seen at 50°C after 15 minutes, and for Epidermophyton floccosum at 50°C after 10 minutes. Limited growth of Scytalidium was seen at 55°C after 5 minutes. No inhibition was observed after laser treatment of fungal colonies or suspensions. In vivo treatment of toenails showed no improvement in Onychomycosis Severity Index score. The Nd:YAG 1064-nm laser was the only laser tested. Laser treatment of onychomycosis was not related to thermal damage or direct laser effects. In vivo treatment did not result in onychomycosis cure.
Sigma-Aldrich
Aluminum, powder, <5 μm particle size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Sigma-Aldrich
Neodymium, powder, −40 mesh, ≥99% trace rare earth metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Aluminum, powder, <75 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, ≤30 μm, spherical powder, ≥99%
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Sigma-Aldrich
Yttrium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Sigma-Aldrich
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Sigma-Aldrich
Yttrium sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis
Aluminum, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, CP