Merck
  • Efficacy and safety of infliximab plus naproxen versus naproxen alone in patients with early, active axial spondyloarthritis: results from the double-blind, placebo-controlled INFAST study, Part 1.
Efficacy and safety of infliximab plus naproxen versus naproxen alone in patients with early, active axial spondyloarthritis: results from the double-blind, placebo-controlled INFAST study, Part 1.
Annals of the rheumatic diseases (2013-05-23)
J Sieper, J Lenaerts, J Wollenhaupt, M Rudwaleit, V I Mazurov, L Myasoutova, S Park, Y Song, R Yao, D Chitkara, N Vastesaeger
ABSTRACT
To assess whether combination therapy with infliximab (IFX) plus nonsteroidal anti-inflammatory drugs (NSAIDs) is superior to NSAID monotherapy for reaching Assessment of SpondyloArthritis international Society (ASAS) partial remission in patients with early, active axial spondyloarthritis (SpA) who were naïve to NSAIDs or received a submaximal dose of NSAIDs. Patients were randomised (2 : 1 ratio) to receive naproxen (NPX) 1000 mg daily plus either IFX 5 mg/kg or placebo (PBO) at weeks 0, 2, 6, 12, 18 and 24. The primary efficacy measure was the percentage of patients who met ASAS partial remission criteria at week 28. Several other measures of disease activity, clinical symptoms and patient-rated outcomes were evaluated. Treatment group differences were analysed with Fisher exact tests or analysis of covariance. A greater percentage of patients achieved ASAS partial remission in the IFX+NPX group (61.9%; 65/105) than in the PBO+NPX group (35.3%; 18/51) at week 28 (p=0.002) and at all other visits (p<0.05, all comparisons). Results of most other disease activity and patient-reported endpoints (including Ankylosing Spondylitis Disease Activity Score, Bath Ankylosing Spondylitis Disease Activity Index, Bath Ankylosing Spondylitis Functional Index, multiple quality of life measures and pain measures) showed greater improvement in the IFX+NPX group than the PBO+NPX group, with several measures demonstrating early and consistent improvement over 28 weeks of treatment. Patients with early, active axial SpA who received IFX+NPX combination treatment were twice as likely to achieve clinical remission as patients who received NPX alone. NPX alone led to clinical remission in a third of patients.
Product Description
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Naproxen, meets USP testing specifications
Supelco
Naproxen, VETRANAL®, analytical standard
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Supelco
Naproxen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Naproxen, European Pharmacopoeia (EP) Reference Standard