Merck
  • Ultrasound-assisted oxidative desulfurization of bunker-C oil using tert-butyl hydroperoxide.
Ultrasound-assisted oxidative desulfurization of bunker-C oil using tert-butyl hydroperoxide.
Ultrasonics sonochemistry (2013-03-30)
Qiong Tang, Song Lin, Ying Cheng, Sujun Liu, Jun-Ru Xiong
ABSTRACT
This work investigated the ultrasonic assisted oxidative desulfurization of bunker-C oil with TBHP/MoO3 system. The operational parameters for the desulfurization procedure such as ultrasonic irradiation time, ultrasonic wave amplitude, catalyst initial concentration and oxidation agent initial concentration were studied. The experimental results show that the present oxidation system was very efficient for the desulfurization of bunker-C oil and ~35% sulfur was removed which was dependent on operational parameters. The application of ultrasonic irradiation allowed sulfur removal in a shorter time. The stronger the solvent polarity is, the higher the sulfur removal rate, but the recovery rate of oil is lower. The sulfur compounds in bunker-C oil reacted with TBHP to produce corresponding sulfoxide, and further oxidation produced the corresponding sulfone.
Sigma-Aldrich
tert-Butyl hydroperoxide solution, CP
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in nonane
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane