Merck
  • CoOOH nanosheet electrodes: Simple fabrication for sensitive electrochemical sensing of hydrogen peroxide and hydrazine.
CoOOH nanosheet electrodes: Simple fabrication for sensitive electrochemical sensing of hydrogen peroxide and hydrazine.
Biosensors & bioelectronics (2012-08-18)
Kian Keat Lee, Pui Yee Loh, Chorng Haur Sow, Wee Shong Chin
ABSTRACT
Cobalt oxyhydroxide, CoOOH, nanosheets were prepared via a surface alkaline treatment of cobalt foil at room temperature without using templates and catalysts. The morphology, chemical composition and structures of the nanosheets were characterized by XRD, FTIR and Raman spectroscopy, FESEM and TEM. These oriented and nanostructured arrays can be used directly as electrodes, thus simplifying the electrode fabrication process, as well as offering advantages such as enhanced electrode-electrolyte contact area, minimum diffusion resistance and direct active material-current collector connection for fast electron transport. The electrode was used as an electrochemical sensor towards non-enzymatic detection of hydrogen peroxide and hydrazine in alkaline solution. The amperometric detection of H(2)O(2) and N(2)H(4) was carried out at low potential (0V and 0.1V). At 0.1V, the amperometric signals are linearly proportional to H(2)O(2) concentration up to 1.6mM (R(2)=0.995), showing a detection limit (S/N=3) of 40μM and a high sensitivity of 99μA mM(-1)cm(-2). For N(2)H(4), the amperometric signals are linearly proportional to concentration up to 1.2mM (R(2)=0.99), showing a detection limit (S/N=3) of 20μM and a high sensitivity of 155μA mM(-1)cm(-2) at 0.1V.
Product Description
Cobalt, foil, not light tested, 50x50mm, thickness 0.02mm, 99.9%
Cobalt, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.007mm, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.05mm, as rolled, 99.9%
Cobalt, foil, light tested, 25x25mm, thickness 0.025mm, 99.9%
Cobalt, foil, not light tested, 150x150mm, thickness 0.0125mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.01mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.009mm, 99.9%
Cobalt, foil, light tested, 50x50mm, thickness 0.01mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.007mm, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.004mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.004mm, 99.9%
Cobalt, foil, not light tested, 150x150mm, thickness 0.01mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.025mm, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.005mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.008mm, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.9%
Cobalt, foil, light tested, 25x25mm, thickness 0.02mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.002mm, temporary acrylic support, 99.9%
Cobalt, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.9%
Cobalt, foil, light tested, 50x50mm, thickness 0.0125mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.01mm, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.009mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.015mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.0025mm, temporary acrylic support, 99.9%
Cobalt, foil, light tested, 50x50mm, thickness 0.015mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.02mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.003mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.006mm, 99.9%