Merck
Hydrothermally Grown ZnO Micro/Nanotube Arrays and Their Properties.
Nanoscale research letters (2009-01-01)
Huibo Chen, Xiang Wu, Lihong Gong, Cai Ye, Fengyu Qu, Guozhen Shen
ABSTRACT
We reported the optical and wettability properties of aligned zinc oxide micro/nanotube arrays, which were synthesized on zinc foil via a simple hydrothermal method. As-synthesized ZnO micro/nanotubes have uniform growth directions along the [0001] orientations with diameters in the range of 100-700 nm. These micro/nanotubes showed a strong emission peak at 387 nm and two weak emission peaks at 422 and 485 nm, respectively, and have the hydrophobic properties with a contact angle of 121°. Single ZnO micro/nanotube-based field-effect transistor was also fabricated, which shows typical n-type semiconducting behavior.
Product Description
Zinc, foil, 25mm disks, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 1m coil, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 50mm disks, thickness 0.125mm, as rolled, 99.99+%
Zinc, foil, 50mm disks, thickness 0.5mm, as rolled, 99.95+%
Zinc, foil, 50mm disks, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 50mm disks, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 4mm disks, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 25mm disks, thickness 1.0mm, as rolled, 99.95+%
Zinc, foil, 50mm disks, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 25mm disks, thickness 1.0mm, as rolled, 99.99+%
Zinc, foil, 4mm disks, thickness 0.25mm, as rolled, 99.95+%
Zinc, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.99+%
Zinc, foil, 50mm disks, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 0.5mm, as rolled, 99.95+%
Zinc, foil, 5m coil, thickness 0.075mm, as rolled, 99.95+%
Zinc, foil, 4mm disks, thickness 0.01mm, 99.9%
Zinc, foil, 6mm disks, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 2m coil, thickness 1.0mm, as rolled, 99.95+%
Zinc, foil, 4mm disks, thickness 0.015mm, 99.9%
Zinc, foil, 6mm disks, thickness 0.005mm, 99.9%
Zinc, foil, 2m coil, thickness 0.075mm, as rolled, 99.95+%
Zinc, foil, 8mm disks, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 50mm disks, thickness 0.25mm, as rolled, 99.99+%
Zinc, foil, not light tested, 25x25mm, thickness 0.005mm, 99.9%
Zinc, foil, 4mm disks, thickness 0.5mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 0.0025mm, 99.9%
Zinc, foil, 8mm disks, thickness 0.02mm, 99.9%
Zinc, foil, 2m coil, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%
Zinc, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.95+%