Merck
Cross-medal arrays of Ta-doped rutile titania.
Journal of the American Chemical Society (2009-08-27)
Xianfeng Yang, Jian Chen, Li Gong, Mingmei Wu, Jimmy C Yu
ABSTRACT
A rutile microstructure with a novel cross-medal morphology was prepared via a hydrothermal method by using a Tantalum foil as both the doping source and the substrate. This rutile cross-medal prefers to grow along an unprecedented preferential orientation of <110> directions from the substrate, which is very different from the traditional c-axis. The new material exhibits highly enhanced field emission (FE) properties.
Product Description
Tantalum, foil, light tested, 50x50mm, thickness 0.01mm, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.01mm, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.075mm, annealed, 99.9%
Tantalum, foil, light tested, 100x100mm, thickness 0.05mm, annealed, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.01mm, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.001mm, 99.9%
Tantalum, foil, 15mm disks, thickness 0.20mm, annealed, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.003mm, 99.9%
Tantalum, foil, 1m coil, thickness 0.025mm, as rolled, 99.9%
Tantalum, foil, light tested, 150x300mm, thickness 0.075mm, annealed, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.004mm, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.034mm, as rolled, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.02mm, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.006mm, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.01mm, 99.9%
Tantalum, foil, not light tested, 100x100mm, thickness 0.009mm, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.0015mm, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.003mm, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.004mm, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.006mm, 99.9%
Tantalum, foil, light tested, 150x150mm, thickness 0.03mm, as rolled, 99.9%
Tantalum, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.0015mm, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.003mm, 99.9%
Tantalum, foil, not light tested, 82x100mm, thickness 0.006mm, 99.9%
Tantalum, foil, not light tested, 100x100mm, thickness 0.003mm, 99.9%
Tantalum, foil, 10mm disks, thickness 0.03mm, as rolled, 99.9%
Tantalum, foil, 10mm disks, thickness 0.0075mm, 99.9%
Tantalum, foil, 10mm disks, thickness 0.0125mm, 99.9%
Tantalum, foil, 100x100mm, thickness 0.20mm, annealed, 99.9%