- Alternative methods for the production of carrier-free (66,67)Ga.
Alternative methods for the production of carrier-free (66,67)Ga.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2001-02-24)
D Nayak, S Lahiri
PMID11200879
ABSTRACT
Heavy ion activation of natural cobalt foil with 52 MeV 11B4+ and 73 MeV 12C6+ results in the formation of carrier-free (66,67)Ge and (66,67)As radionuclides, respectively, along with their corresponding decay products, (66,67)Ga in the matrix. The carrier free gallium radionuclides have been separated from the bulk target matrix cobalt by employing liquid-liquid extraction method using TOA as liquid anion exchanger.
Product Description
Cobalt, foil, 15mm disks, thickness 0.125mm, as rolled, 99.99+%
Cobalt, foil, not light tested, 50x50mm, thickness 0.02mm, 99.9%
Cobalt, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.007mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.05mm, as rolled, 99.9%
Cobalt, foil, light tested, 25x25mm, thickness 0.01mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.125mm, as rolled, 99.99+%
Cobalt, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.01mm, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.009mm, 99.9%
Cobalt, foil, light tested, 25x25mm, thickness 0.025mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.0025mm, temporary acrylic support, 99.9%
Cobalt, foil, light tested, 50x50mm, thickness 0.015mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.25mm, as rolled, 99.9%
Cobalt, foil, 8mm disks, thickness 0.0125mm, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.02mm, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.002mm, temporary acrylic support, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.0025mm, temporary acrylic support, 99.9%
Cobalt, foil, 8mm disks, thickness 0.015mm, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.004mm, 99.9%
Cobalt, foil, light tested, 50x50mm, thickness 0.0125mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.25mm, as rolled, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.009mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.007mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.01mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.015mm, 99.9%
Cobalt, foil, 50x50mm, thickness 1.0mm, as rolled, 99.9%
Cobalt, foil, 10mm disks, thickness 0.006mm, 99.9%
Cobalt, foil, 15mm disks, thickness 0.05mm, annealed, 99.9%