Merck
Benzoyl peroxide: percutaneous penetration and metabolic disposition.
Journal of the American Academy of Dermatology (1981-01-01)
S Nacht, D Yeung, J N Beasley, M D Anjo, H I Maibach
ABSTRACT
The transepidermal penetration and metabolic disposition of 14C-benzoyl peroxide were assessed in vitro (excised human skin) and in vivo (rhesus monkey). In vitro, the benzoyl peroxide penetrated into the skin, through the stratum corneum or the follicular openings, or both, and was recovered on the dermal side as benzoic acid. In vivo, benzoic acid was recovered from urine in amounts equivalent to 45% and 98% of the radiolabel following, respectively, topical and intramuscular administration of small amounts of 14C-benzoyl peroxide. We conclude that benzoyl peroxide penetrates as such into the skin layers and is converted therein to benzoic acid, which, in turn is absorbed into the systemic circulation. Renal clearance of the metabolite is sufficiently rapid as to preclude its hepatic conjugation with glycine, since following topical administration to rhesus monkeys, no hippuric acid was found in the urine, as could have been expected had a significant amount of benzoic acid passed through the liver.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzoic acid, SAJ first grade, ≥99.5%
Sigma-Aldrich
Benzoic acid, SAJ special grade, ≥99.5%
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland