Merck
  • In vitro iontophoresis of R-apomorphine across human stratum corneum. Structure-transport relationship of penetration enhancement.
In vitro iontophoresis of R-apomorphine across human stratum corneum. Structure-transport relationship of penetration enhancement.
Journal of controlled release : official journal of the Controlled Release Society (2002-10-26)
Gai Ling Li, Ronald van der Geest, Laurence Chanet, Elske van Zanten, Meindert Danhof, Joke A Bouwstra
ABSTRACT
To achieve a therapeutical effect of the anti-Parkinson's drug R-apomorphine via iontophoresis delivery, enhancement strategies in vitro were explored using three structurally related enhancers, lauric acid (LA), dodecyltrimethylammonium bromide (DTAB) and Laureth-3 oxyethylene ether (C(12)EO(3)). Human stratum corneum and shed snake skin were pretreated with 0.15 M each enhancer solution in propylene glycol (PG). Thereafter, passive diffusion, iontophoretic transport and post-iontophoretic passive diffusion were investigated. Compared to the control (PG pretreatment), a slight inhibition on both passive and iontophoretic delivery was observed with cationic surfactant DTAB pretreated stratum corneum. Pretreatment with anionic surfactant LA resulted in a great enhancement on passive delivery, but only a small enhancing effect on the iontophoretic delivery. Unlike the others, the nonionic surfactant C(12)EO(3) substantially increased iontophoretic transport rate of R-apomorphine by 2.3-fold, whereas passive delivery was basically unchanged or slightly affected. The magnitude of enhancing effect of C(12)EO(3) was dependent on the surfactant concentration and the pretreatment duration. Moreover, comparison of transport data through shed snake skin with human stratum corneum indicates that both shunt- and intercellular pathways are involved in the iontophoretic transport of R-apomorphine.
Sigma-Aldrich
1,2-Propanediol, SAJ first grade, ≥99.0%
Sigma-Aldrich
1,2-Propanediol, SAJ special grade, ≥99.0%
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material