Merck
  • Evaluation of the health aspects of methyl paraben: a review of the published literature.
Evaluation of the health aspects of methyl paraben: a review of the published literature.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2002-10-22)
M G Soni, S L Taylor, N A Greenberg, G A Burdock
ABSTRACT
Methyl paraben (CAS No. 99-76-3) is a methyl ester of p-hydroxybenzoic acid. It is a stable, non-volatile compound used as an antimicrobial preservative in foods, drugs and cosmetics for over 50 years. Methyl paraben is readily and completely absorbed through the skin and from the gastrointestinal tract. It is hydrolyzed to p-hydroxybenzoic acid, conjugated, and the conjugates are rapidly excreted in the urine. There is no evidence of accumulation. Acute toxicity studies in animals indicate that methyl paraben is practically non-toxic by both oral and parenteral routes. In a population with normal skin, methyl paraben is practically non-irritating and non-sensitizing. In chronic administration studies, no-observed-effect levels (NOEL) as high as 1050 mg/kg have been reported and a no-observed-adverse-effect level (NOAEL) in the rat of 5700 mg/kg is posited. Methyl paraben is not carcinogenic or mutagenic. It is not teratogenic or embryotoxic and is negative in the uterotrophic assay. The mechanism of cytotoxic action of parabens may be linked to mitochondrial failure dependent on induction of membrane permeability transition accompanied by the mitochondrial depolarization and depletion of cellular ATP through uncoupling of oxidative phosphorylation. Parabens are reported to cause contact dermatitis reactions in some individuals on cutaneous exposure. Parabens have been implicated in numerous cases of contact sensitivity associated with cutaneous exposure; however, the mechanism of this sensitivity is unknown. Sensitization has occurred when medications containing parabens have been applied to damaged or broken skin. Allergic reactions to ingested parabens have been reported, although rigorous evidence of the allergenicity of ingested paraben is lacking.
Product Description
Supelco
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Methyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
USP
Methylparaben, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
Methyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Supelco
4-Hydroxybenzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methyl 4-hydroxybenzoate, SAJ first grade, ≥98.0%