Merck
  • Discrimination of complete hydatidiform mole from its mimics by immunohistochemistry of the paternally imprinted gene product p57KIP2.
Discrimination of complete hydatidiform mole from its mimics by immunohistochemistry of the paternally imprinted gene product p57KIP2.
The American journal of surgical pathology (2001-11-02)
D H Castrillon, D Sun, S Weremowicz, R A Fisher, C P Crum, D R Genest
ABSTRACT
The p57KIP2 protein is a cell cycle inhibitor and tumor suppressor encoded by a strongly paternally imprinted gene. We explored the utility of p57KIP2 as a diagnostic marker in hydatidiform mole, a disease likely the result of abnormal dosage and consequent misexpression of imprinted genes. Using a monoclonal antibody on paraffin-embedded, formalin-fixed tissue sections, the authors evaluated p57KIP2 expression in normal placenta and in 149 gestations including 59 complete hydatidiform moles, 39 PHMs, and 51 spontaneous losses with hydropic changes. p57KIP2 was strongly expressed in cytotrophoblast and villous mesenchyme in normal placenta, all cases of partial hydatidiform moles (39 of 39) and all spontaneous losses with hydropic changes (51 of 51). In contrast, p57KIP2 expression in cytotrophoblast and villous mesenchyme was absent or markedly decreased in 58 of 59 complete hydatidiform moles. In all gestations p57KIP2 was strongly expressed in decidua and in intervillous trophoblast islands, which served as internal positive controls for p57KIP2 immunostaining. p57KIP2 immunohistochemistry can reliably identify most cases of complete hydatidiform mole irrespective of gestational age and is thus a useful diagnostic adjunct, complementary to ploidy analysis, in the diagnosis of hydatidiform mole.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde neutral buffer solution, 7.5% formaldehyde in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, 10%
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde neutral buffer solution, 3.7% formaldehyde in H2O
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.