Merck
  • Analgesic and immunomodulatory effects of codeine and codeine 6-glucuronide.
Analgesic and immunomodulatory effects of codeine and codeine 6-glucuronide.
Pharmaceutical research (1996-02-01)
V Srinivasan, D Wielbo, J Simpkins, J Karlix, K Sloan, I Tebbett
ABSTRACT
The antinociceptive and immunosuppressive effects of codeine and codeine 6-glucuronide were determined in rats after intracerebroventricular administration. Codeine 6-glucuronide was synthesized using a modification of the Koenigs-Knorr reaction. A lipophilic intermediate formed during synthesis, methyl [codein-6-yl-2,3,4-tri-O-acetyl-beta-D-glucopyranosid] uronate, was also tested. Morphine was used as a positive control to compare antinociceptive potencies of these compounds. All compounds tested produced significant analgesic responses, as assessed by the tail flick model. Additionally, codeine 6-glucuronide showed significantly less immunosuppressive effects than codeine in vitro. We conclude that codeine 6-glucuronide and related compounds may have clinical benefit in the treatment of pain in immune compromised patients.
Product Description
Supelco
Codeine-6-β-D-glucuronide solution, 100 μg/mL in methanol: water (2:8), ampule of 1 mL, certified reference material, Cerilliant®