Merck
  • Conversion of xylan, d-xylose and lignocellulosic biomass into furfural using AlCl3 as catalyst in ionic liquid.
Conversion of xylan, d-xylose and lignocellulosic biomass into furfural using AlCl3 as catalyst in ionic liquid.
Bioresource technology (2013-01-12)
Luxin Zhang, Hongbing Yu, Pan Wang, Heng Dong, Xinhong Peng
ABSTRACT
In order to define a new green catalytic pathway for the production of furfural, the catalyzed conversion of xylan into furfural in 1-butyl-3-methylimidazolium chloride was studied by using mineral acids and metal chlorides as catalysts under microwave irradiation. Amongst these catalysts, AlCl(3) resulted in the highest furfural yield of 84.8% at 170°C for 10s. The effect of AlCl(3) on the conversion efficiency of d-xylose and untreated lignocellulosic biomass was also investigated, the yields of furfural from corncob, grass and pine wood catalyzed by AlCl(3) in [BMIM]Cl were in the range of 16-33%. [BMIM]Cl and AlCl(3) could be recycled for four runs with stable catalytic activity. AlCl(3) is less corrosive than mineral acids, and the use of ionic liquid as reaction medium will no longer generate toxic wastewater, thus this reaction system is more ecologically viable.
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
Sigma-Aldrich
Furfural, natural, ≥98%, FCC, FG
Sigma-Aldrich
Aluminum chloride, SAJ first grade, ≥98.0%
Sigma-Aldrich
Furfural, SAJ first grade, ≥99.0%
Sigma-Aldrich
Furfural, ≥98%, FCC, FG
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Furfural, 99%
Sigma-Aldrich
Furfural, ACS reagent, 99%
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, ReagentPlus®, 99%
Sigma-Aldrich
Aluminum chloride, anhydrous, sublimed, ≥98%
Sigma-Aldrich
Aluminum chloride solution, 1.0 M in nitrobenzene
Sigma-Aldrich
Aluminum chloride, reagent grade, 98%
Sigma-Aldrich
Aluminum chloride hydrate, 99.999% trace metals basis