Merck
  • Ab initio structure modelling of complex thin-film oxides: thermodynamical stability of TiC/thin-film alumina.
Ab initio structure modelling of complex thin-film oxides: thermodynamical stability of TiC/thin-film alumina.
Journal of physics. Condensed matter : an Institute of Physics journal (2010-01-13)
J Rohrer, C Ruberto, P Hyldgaard
ABSTRACT
We present a strategy to identify energetically favourable oxide structures in thin-film geometries. Thin-film candidate configurations are constructed from a pool of sublattices of stable and metastable oxide bulk phases. Favourable stoichiometric compositions and atomic geometries are identified by comparing total and Gibbs free energies of the relaxed configurations. This strategy is illustrated for thin-film alumina on TiC, materials which are commonly fabricated by chemical vapour deposition (CVD) and used as wear-resistant multilayer coatings. Based on the standard implementation of ab initio thermodynamics, with an assumption of equilibrium between molecular O(2) and the oxide, we predict a stability preference of TiC/alumina configurations that show no binding across the interface. This result is seemingly in conflict with the wear-resistant character of the material and points towards a need for extending standard ab initio thermodynamics to account for relevant growth environments.
Sigma-Aldrich
Titanium carbide, nanopowder, ≤200 nm particle size (SEM)
Sigma-Aldrich
Titanium(IV) carbide, −325 mesh, 98%
Sigma-Aldrich
Titanium(IV) carbide, powder, <4 μm, ≥99% (Ti)