Merck
  • Preparation and characterization of silver-doped nanobioactive glass particles and their in vitro behaviour for biomedical applications.
Preparation and characterization of silver-doped nanobioactive glass particles and their in vitro behaviour for biomedical applications.
Journal of nanoscience and nanotechnology (2013-07-26)
Muthusamy Prabhu, Kandiah Kavitha, Rangaraj Suriyaprabha, Palanisamy Manivasakan, Venkatachalam Rajendran, Palanisami Kulandaivelu
ABSTRACT
In this study, silver-doped silica- and phosphate-based nanobioactive glass compositions (58SiO2-(33- x)CaO-9P2O5-xAg2O) (x = 0, 0.5, 1, 2 and 3 mol%) were synthesised by a simple and cost-effective sol-gel method. The prepared samples were characterised by X-ray diffraction, Fourier transform infrared spectroscopy, transmission electron microscopy, scanning electron microscopy and energy-dispersive X-ray fluorescence spectrometer studies. All the compositions of the glass samples revealed amorphous phase with spherical morphology and a particle size less than 100 nm. The prepared glass samples reveal the specific surface area in the range of 55.31-90.69 m2 g(-1). The bioactivity of glass samples was confirmed through the formation of the hydroxyapatite layer on glass surfaces during in vitro studies in which silver doped glasses (2 and 3 mol%) showed better bioactivity. A better biocompatibility was achieved in human gastric adenocarcinoma cell line in case of silver-free glass sample while comparing the biological behaviour of Ag2O-doped glasses. Further, the Ag2O-doped nanobioactive glasses revealed significant antibacterial activity against Staphylococcus aureus and Escherichia coli. Ag2O substitutions showed better in vitro bioactivity and remained slightly toxic to human cells at a concentration of 100 microg mL(-1). Silver-doped nanobioactive glass shows good antimicrobial property as well as no significant toxic for implant applications.
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Silver nanowires, diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver nanowires, diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis