Merck
  • Microalgae mediated synthesis of silver nanoparticles and their antibacterial activity against pathogenic bacteria.
Microalgae mediated synthesis of silver nanoparticles and their antibacterial activity against pathogenic bacteria.
Indian journal of experimental biology (2013-07-05)
S S Sudha, Karthic Rajamanickam, J Rengaramanujam
ABSTRACT
Silver nanoparticles is known to have antimicrobial affects. Cyanobacteria isolates from muthupet mangrove includes Aphanothece sp, Oscillatoria sp, Microcoleus sp, Aphanocapsa sp, Phormidium sp, Lyngbya sp, Gleocapsa sp, Synechococcus sp, Spirulina sp with were set in compliance with their cellular mechanism of nano silver creation, and were investigated by UV-VIS spectrophotometer, Energy-dispersive X-ray (EDX) and scanning electron microscopy (SEM). Silver nanoparticles were spherical shaped well distributed without aggregation in solution with an average size of about 40- 80 nm. Synthesised nano silver had antibacterial production on various organisms that provoked various diseases in humans. The cellular metabolites of Microcoleus sp. only created nano silver and it enhanced the antibacterial activity against test pathogenic bacteria from MTCC (Proteus vulgaris, Salmonella typhi, Vibrio cholera, Streptococcus sp., Bacillus subtilis, Staphylococcus aureus, Escherichia coli.) The antimicrobial assay was performed using 0.001 M concentration of nano silver in well diffusion method with positive control of appropriate standard antibiotic discs Cephotaxime, Ampicillin, Tetracyclin, Cephalexin etc. Synthesised silver nanoparticles acted as an effective antimicrobial agent and proved as an alternative for the development of new antimicrobial agents to combat the problem of resistance.
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Silver nanowires, diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver nanowires, diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis